Search
Search

ZNF436-AS1| Gene Symbol: ZNF436-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033690.1 | 1 | 432 | |||
| NR_033691.1 | 1 | 185 | ||||
| GenBank mRNA | AK074989.1 | - | 1 | 346 | ||
| BC028411.1 | - | 1 | 173 | 4822173 | ||
| BC054115.1 | - | 1 | 436 | 5270027 | ||
| BC068594.1 | - | 1 | 414 | 4828713 | ||
| BQ072309.1 | - | 420 | ||||
| BX098506.1 | - | 1 | 153 | |||