Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TP53TG1 SIRNA Assays ›
          Gene Symbol:
          TP53TG1
          Gene Name:
          TP53 target 1 (non-protein coding)
          Gene Aliases:
          LINC00096, NCRNA00096, P53TG1, P53TG1-D, TP53AP1
          Chromosome Location:
          Chr.7: 87325347 - 87345492 on Build GRCh38
          UniGene ID:
          Hs.274329
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n543785

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TP53TG1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_015381.1 2 530
          GenBank mRNA AB007455.1 - 2 553
          AB007456.1 - 3 482
          AB007457.1 - 2 333
          AB007458.1 - 2 262
          AK292782.1 - 2 547
          BC002709.2 - 2 530
          BC061927.1 - 2 483
          BC103631.1 - 2 237
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BI258711.1 - 2 541 Chr. 7 - 87325362 - 87345491 [-] on Build GRCh38
          GenBankmRNA BM912218.1 - 465 Chr. - - [-] on Build
          GenBankmRNA BP318778.1 - 2 542 Chr. 7 - 87341560 - 87345505 [-] on Build GRCh38
          GenBankmRNA BE393755.1 - 2 511 Chr. 7 - 87341517 - 87345473 [-] on Build GRCh38
          GenBankmRNA CF124441.1 - 2 390 Chr. 7 - 87341517 - 87345314 [-] on Build GRCh38
          GenBankmRNA DA741956.1 - 3 471 Chr. 7 - 87341555 - 87345504 [-] on Build GRCh38
          GenBankmRNA CN409086.1 - 2 506 Chr. 7 - 87341578 - 87345468 [-] on Build GRCh38
          GenBankmRNA DB202605.1 - 2 547 Chr. 7 - 87341568 - 87345509 [-] on Build GRCh38
          GenBankmRNA BM850328.1 - 2 226 Chr. 7 - 87325370 - 87345188 [-] on Build GRCh38
          GenBankmRNA BG033277.1 - 500 Chr. - - [-] on Build
          GenBankmRNA BM912196.1 - 465 Chr. - - [-] on Build
          GenBankmRNA N47733.1 - 2 120 Chr. 7 - 87325237 - 87345082 [-] on Build GRCh38
          GenBankmRNA BM915584.1 - 2 465 Chr. 7 - 87325297 - 87345427 [-] on Build GRCh38
          GenBankmRNA BQ439368.1 - 2 537 Chr. 7 - 87325355 - 87345499 [-] on Build GRCh38
          GenBankmRNA BG831472.1 - 2 541 Chr. 7 - 87325297 - 87345501 [-] on Build GRCh38
          GenBankmRNA CN409085.1 - 2 511 Chr. 7 - 87325365 - 87345473 [-] on Build GRCh38
          GenBankmRNA BM554619.1 - 482 Chr. - - [-] on Build
          GenBankmRNA AA447887.1 - 2 313 Chr. 7 - 87325353 - 87345275 [-] on Build GRCh38
          GenBankmRNA BQ961841.1 - 2 538 Chr. 7 - 87325297 - 87345500 [-] on Build GRCh38
          GenBankmRNA BQ581775.1 - 2 452 Chr. 7 - 87325398 - 87345407 [-] on Build GRCh38
          GenBankmRNA BM550903.1 - 2 465 Chr. 7 - 87325297 - 87345419 [-] on Build GRCh38
          GenBankmRNA BU536344.1 - 2 242 Chr. 7 - 87325347 - 87345201 [-] on Build GRCh38
          GenBankmRNA CB994267.1 - 2 547 Chr. 7 - 87325360 - 87345482 [-] on Build GRCh38
          GenBankmRNA BM550333.1 - 2 466 Chr. 7 - 87325261 - 87345428 [-] on Build GRCh38
          GenBankmRNA BX110032.1 - 2 497 Chr. 7 - 87325347 - 87345459 [-] on Build GRCh38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          response to stress
          cellular response to DNA damage stimulus
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0