Search
Search

ZFHX4-AS1| Gene Symbol: ZFHX4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024360.1 | 4 | 607 | |||
| GenBank mRNA | BC037827.1 | - | 4 | 615 | ||
| DA140947.1 | - | 4 | 504 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG699142.1 | - | 4 | 615 | Chr. 8 - 76611813 - 76683278 [-] on Build GRCh38 | |