Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HAND2-AS1 SIRNA Assays ›
          Gene Symbol:
          HAND2-AS1
          Gene Name:
          HAND2 antisense RNA 1 (head to head)
          Gene Aliases:
          DEIN, NBLA00301
          Chromosome Location:
          Chr.4: 173528600 - 173591255 on Build GRCh38
          UniGene ID:
          Hs.61435
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n544250

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HAND2-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003679.2 1 62
          NR_136192.1 1 149
          NR_136193.1 1 149
          NR_136194.1 1 149
          NR_136195.1 1 149
          NR_136196.1 1 149
          NR_136198.1 1 149
          NR_136199.1 1 149
          NR_136200.1 1 149
          NR_136201.1 1 149
          GenBank mRNA AB075501.1 - 1 61
          AK054605.1 - 1 58
          AU253586.1 - 1 58
          BG519698.1 - 1 28
          BQ882603.1 - 1 149
          DA665500.1 - 1 58
          DB292439.1 - 1 82
          DC386781.1 - 1 60
          EF529495.1 - 1 62
          EF529496.1 - 1 62
          EF529497.1 - 1 62
          EF529498.1 - 1 62
          EF529499.1 - 1 62
          EF529500.1 - 1 62
          EF529501.1 - 1 62
          EF529502.1 - 1 62
          EF529503.1 - 1 62
          EF529504.1 - 1 62
          EF529505.1 - 1 62
          EF529506.1 - 1 62
          EF529507.1 - 1 62
          EF529508.1 - 1 62
          EF529509.1 - 1 62
          EU000853.1 - 1 62
          EU000854.1 - 1 62
          EU000855.1 - 1 62
          EU000856.1 - 1 62
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DC360108.1 - 1 49 Chr. 4 - 173530472 - 173538277 [+] on Build GRCh38
          GenBankmRNA DC386933.1 - 1 45 Chr. 4 - 173530475 - 173538159 [+] on Build GRCh38
          GenBankmRNA DA614232.1 - 1 58 Chr. 4 - 173530462 - 173537219 [+] on Build GRCh38
          GenBankmRNA BF304375.1 - 38 Chr. - - [-] on Build
          GenBankmRNA DC354299.1 - 1 58 Chr. 4 - 173530462 - 173538157 [+] on Build GRCh38
          GenBankmRNA DC360375.1 - 1 60 Chr. 4 - 173530460 - 173538107 [+] on Build GRCh38
          GenBankmRNA DC360232.1 - 1 30 Chr. 4 - 173530491 - 173538106 [+] on Build GRCh38
          GenBankmRNA DA927156.1 - 1 58 Chr. 4 - 173530462 - 173537450 [+] on Build GRCh38
          GenBankmRNA BE312774.1 - 1 18 Chr. 4 - 173530502 - 173537295 [+] on Build GRCh38
          GenBankmRNA BP379670.1 - 1 44 Chr. 4 - 173530475 - 173538344 [+] on Build GRCh38
          GenBankmRNA DA919798.1 - 1 58 Chr. 4 - 173530462 - 173537054 [+] on Build GRCh38
          GenBankmRNA DC386963.1 - 1 25 Chr. 4 - 173530495 - 173538165 [+] on Build GRCh38
          GenBankmRNA DA606691.1 - 1 24 Chr. 4 - 173530496 - 173537315 [+] on Build GRCh38
          GenBankmRNA DC360466.1 - 1 58 Chr. 4 - 173530462 - 173538130 [+] on Build GRCh38
          GenBankmRNA DC297746.1 - 1 51 Chr. 4 - 173530469 - 173538168 [+] on Build GRCh38
          GenBankmRNA DC381495.1 - 1 60 Chr. 4 - 173530460 - 173538307 [+] on Build GRCh38
          GenBankmRNA DC381155.1 - 1 45 Chr. 4 - 173530475 - 173538158 [+] on Build GRCh38
          GenBankmRNA DC360477.1 - 1 45 Chr. 4 - 173530475 - 173538162 [+] on Build GRCh38
          GenBankmRNA BQ777900.1 - 1 130 Chr. 4 - 173530390 - 173538233 [+] on Build GRCh38
          GenBankmRNA DC388584.1 - 1 58 Chr. 4 - 173530462 - 173538318 [+] on Build GRCh38
          GenBankmRNA R24426.1 - 26 Chr. - - [-] on Build
          GenBankmRNA DA823966.1 - 1 45 Chr. 4 - 173530475 - 173537465 [+] on Build GRCh38
          GenBankmRNA DC361029.1 - 1 46 Chr. 4 - 173530475 - 173538187 [+] on Build GRCh38
          GenBankmRNA AL523783.3 - 1 41 Chr. 4 - 173530479 - 173538777 [+] on Build GRCh38
          GenBankmRNA AU252421.1 - 1 87 Chr. 4 - 173530462 - 173537325 [+] on Build GRCh38
          GenBankmRNA AL522836.3 - 1 46 Chr. 4 - 173530474 - 173537415 [+] on Build GRCh38
          GenBankmRNA CK902262.1 - 1 111 Chr. 4 - 173530409 - 173538202 [+] on Build GRCh38
          GenBankmRNA BP380043.1 - 1 60 Chr. 4 - 173530460 - 173538314 [+] on Build GRCh38
          GenBankmRNA DB249244.1 - 1 58 Chr. 4 - 173530462 - 173537046 [+] on Build GRCh38
          GenBankmRNA BX345458.2 - 1 40 Chr. 4 - 173530483 - 173583096 [+] on Build GRCh38
          GenBankmRNA DC360095.1 - 1 58 Chr. 4 - 173530462 - 173538260 [+] on Build GRCh38
          GenBankmRNA DC360996.1 - 1 58 Chr. 4 - 173530462 - 173538121 [+] on Build GRCh38
          GenBankmRNA DC360499.1 - 1 58 Chr. 4 - 173530462 - 173538149 [+] on Build GRCh38
          GenBankmRNA DC354261.1 - 1 58 Chr. 4 - 173530462 - 173538142 [+] on Build GRCh38
          GenBankmRNA DC360579.1 - 1 25 Chr. 4 - 173530495 - 173538334 [+] on Build GRCh38
          GenBankmRNA DA922612.1 - 1 60 Chr. 4 - 173530461 - 173537218 [+] on Build GRCh38
          GenBankmRNA DC380727.1 - 1 58 Chr. 4 - 173530462 - 173538322 [+] on Build GRCh38
          GenBankmRNA BU146691.1 - 1 41 Chr. 4 - 173530479 - 173538629 [+] on Build GRCh38
          GenBankmRNA DC361115.1 - 1 62 Chr. 4 - 173530458 - 173538164 [+] on Build GRCh38
          GenBankmRNA DC360170.1 - 1 58 Chr. 4 - 173530462 - 173538097 [+] on Build GRCh38
          GenBankmRNA DC298067.1 - 1 60 Chr. 4 - 173530460 - 173538137 [+] on Build GRCh38
          GenBankmRNA DC361312.1 - 1 58 Chr. 4 - 173530462 - 173538169 [+] on Build GRCh38
          GenBankmRNA DC380045.1 - 1 58 Chr. 4 - 173530462 - 173538250 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0