Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MEG3 SIRNA Assays ›
          Gene Symbol:
          MEG3
          Gene Name:
          maternally expressed 3 (non-protein coding)
          Gene Aliases:
          FP504, GTL2, LINC00023, NCRNA00023, PRO0518, PRO2160, onco-lncRNA-83, prebp1
          Chromosome Location:
          Chr.14: 100826108 - 100861026 on Build GRCh38
          UniGene ID:
          Hs.654863
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n544288

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MEG3
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002766.2
          5 1089
          NR_003530.2
          7 1339
          NR_003531.3
          6 1206
          NR_033358.1
          6 1219
          NR_033359.1
          6 1198
          NR_033360.1
          5 1105
          NR_046464.1
          6 1153
          NR_046465.2
          5 1127
          NR_046466.1
          4 975
          NR_046467.1
          6 1257
          NR_046469.1
          5 1119
          NR_046470.2
          6 1249
          NR_046471.1
          4 1013
          NR_046472.1
          5 1143
          NR_046473.1
          5 1089
          GenBank mRNA AB032607.1 -
          5 1075
          AF052114.1 -
          3 831
          AJ413186.1 -
          2 2352
          AK055725.1 -
          2 2363
          AK057522.1 -
          4 1225
          AK092504.1 -
          3 1777
          AK092707.1 -
          2 1645
          AK127864.1 -
          2 2952
          AV704361.1 -
          1 13
          AY314975.1 - LIT3321
          MEG3
          6 1205
          BC036882.1 -
          3 214
          BC062783.1 -
          2 541
          BC092509.1 -
          3 284
          GQ183494.1 -
          5 1097
          GQ183495.1 -
          6 1145
          GQ183496.1 -
          5 1120
          GQ183497.1 -
          4 967
          GQ183498.1 -
          6 1249
          GQ183499.1 -
          6 1212
          GQ183500.1 -
          5 1111
          GQ183501.1 -
          6 1242
          GQ183502.1 -
          4 1005
          GQ183503.1 -
          5 1135
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CD513101.1 - 2 166 Chr. 14 - 100835754 - 100861026 [+] on Build GRCh38
          GenBankmRNA BF932705.1 - 333 Chr. - - [-] on Build
          GenBankmRNA BU075874.1 - 2 354 Chr. 14 - 100835573 - 100860746 [+] on Build GRCh38
          GenBankmRNA R81006.1 - 146 Chr. - - [-] on Build
          GenBankmRNA AV701976.1 - 3 239 Chr. 14 - 100835307 - 100860739 [+] on Build GRCh38
          GenBankmRNA BX426730.2 - 2 83 Chr. 14 - 100831491 - 100845533 [+] on Build GRCh38
          GenBankmRNA BX453065.2 - 137 Chr. - - [-] on Build
          GenBankmRNA BX346951.1 - 3 411 Chr. 14 - 100829508 - 100860951 [+] on Build GRCh38
          GenBankmRNA BX408784.2 - 3 328 Chr. 14 - 100829579 - 100861025 [+] on Build GRCh38
          GenBankmRNA AI174720.1 - 3 507 Chr. 14 - 100829397 - 100836304 [+] on Build GRCh38
          GenBankmRNA BM929634.1 - 3 226 Chr. 14 - 100835728 - 100861002 [+] on Build GRCh38
          GenBankmRNA BU786542.1 - 1 404 Chr. 14 - 100835806 - 100860767 [+] on Build GRCh38
          GenBankmRNA BM699430.1 - 4 365 Chr. 14 - 100831460 - 100860756 [+] on Build GRCh38
          GenBankmRNA H59650.1 - 283 Chr. - - [-] on Build
          GenBankmRNA BX408697.2 - 3 279 Chr. 14 - 100829658 - 100861025 [+] on Build GRCh38
          GenBankmRNA BX367746.1 - 3 496 Chr. 14 - 100829409 - 100860888 [+] on Build GRCh38
          GenBankmRNA BU783773.1 - 2 127 Chr. 14 - 100831456 - 100860960 [+] on Build GRCh38
          GenBankmRNA DA361098.1 - 3 268 Chr. 14 - 100829636 - 100845997 [+] on Build GRCh38
          GenBankmRNA AV723467.1 - 3 244 Chr. 14 - 100829658 - 100860731 [+] on Build GRCh38
          GenBankmRNA CB154926.1 - 3 336 Chr. 14 - 100829568 - 100860741 [+] on Build GRCh38
          GenBankmRNA AA029246.1 - 2 126 Chr. 14 - 100831448 - 100860775 [+] on Build GRCh38
          GenBankmRNA BX453064.2 - 2 138 Chr. 14 - 100831437 - 100861026 [+] on Build GRCh38
          GenBankmRNA BX422168.2 - 3 561 Chr. 14 - 100829359 - 100860947 [+] on Build GRCh38
          GenBankmRNA BX408189.1 - 3 442 Chr. 14 - 100829460 - 100860925 [+] on Build GRCh38
          GenBankmRNA AU309923.1 - 3 265 Chr. 14 - 100829639 - 100846016 [+] on Build GRCh38
          GenBankmRNA AI459255.1 - 2 117 Chr. 14 - 100835999 - 100860852 [+] on Build GRCh38
          GenBankmRNA BX423733.2 - 2 84 Chr. 14 - 100831491 - 100861026 [+] on Build GRCh38
          GenBankmRNA BX450687.2 - 3 320 Chr. 14 - 100829584 - 100860988 [+] on Build GRCh38
          GenBankmRNA BI913162.1 - 2 137 Chr. 14 - 100831438 - 100861007 [+] on Build GRCh38
          GenBankmRNA BU579722.1 - 2 97 Chr. 14 - 100831478 - 100860862 [+] on Build GRCh38
          GenBankmRNA BU070003.1 - 1 166 Chr. 14 - 100836048 - 100860862 [+] on Build GRCh38
          GenBankmRNA BX326323.2 - 3 290 Chr. 14 - 100829630 - 100861050 [+] on Build GRCh38
          GenBankmRNA H48704.1 - 2 82 Chr. 14 - 100835837 - 100860825 [+] on Build GRCh38
          GenBankmRNA BY796127.2 - 3 458 Chr. 14 - 100829446 - 100845508 [+] on Build GRCh38
          GenBankmRNA BG703541.1 - 3 289 Chr. 14 - 100829615 - 100846219 [+] on Build GRCh38
          GenBankmRNA CA943189.1 - 3 383 Chr. 14 - 100829522 - 100845520 [+] on Build GRCh38
          GenBankmRNA BU952631.1 - 2 296 Chr. 14 - 100835624 - 100860744 [+] on Build GRCh38
          GenBankmRNA BU787606.1 - 2 97 Chr. 14 - 100831478 - 100861005 [+] on Build GRCh38
          GenBankmRNA BU070059.1 - 1 166 Chr. 14 - 100836048 - 100860906 [+] on Build GRCh38
          GenBankmRNA BX368158.1 - 3 531 Chr. 14 - 100829371 - 100860859 [+] on Build GRCh38
          GenBankmRNA BQ549202.1 - 1 65 Chr. 14 - 100836162 - 100860895 [+] on Build GRCh38
          GenBankmRNA CR741469.1 - 1 130 Chr. 14 - 100836077 - 100861026 [+] on Build GRCh38
          GenBankmRNA BX368151.1 - 5 440 Chr. 14 - 100829640 - 100860985 [+] on Build GRCh38
          GenBankmRNA BX408219.1 - 3 396 Chr. 14 - 100829507 - 100860967 [+] on Build GRCh38
          GenBankmRNA BX450276.1 - 3 420 Chr. 14 - 100829484 - 100846142 [+] on Build GRCh38
          GenBankmRNA BX428872.2 - 2 138 Chr. 14 - 100831437 - 100861026 [+] on Build GRCh38
          GenBankmRNA BX408790.2 - 3 235 Chr. 14 - 100829668 - 100860997 [+] on Build GRCh38
          GenBankmRNA BU070170.1 - 1 166 Chr. 14 - 100836048 - 100860821 [+] on Build GRCh38
          GenBankmRNA BI916294.1 - 3 214 Chr. 14 - 100831481 - 100860983 [+] on Build GRCh38
          GenBankmRNA AA009857.1 - 2 117 Chr. 14 - 100835805 - 100860934 [+] on Build GRCh38
          GenBankmRNA DA083332.1 - 3 391 Chr. 14 - 100829513 - 100836289 [+] on Build GRCh38
          GenBankmRNA BU075431.1 - 3 247 Chr. 14 - 100829679 - 100860862 [+] on Build GRCh38
          GenBankmRNA BM875701.1 - 2 73 Chr. 14 - 100835854 - 100860960 [+] on Build GRCh38
          GenBankmRNA BX429998.2 - 3 214 Chr. 14 - 100829691 - 100846355 [+] on Build GRCh38
          GenBankmRNA BQ268259.1 - 2 141 Chr. 14 - 100835786 - 100860745 [+] on Build GRCh38
          GenBankmRNA BQ669975.1 - 149 Chr. - - [-] on Build
          GenBankmRNA AV727351.1 - 4 525 Chr. 14 - 100829325 - 100845483 [+] on Build GRCh38
          GenBankmRNA AI133465.1 - 2 198 Chr. 14 - 100835722 - 100861003 [+] on Build GRCh38
          GenBankmRNA BX429896.2 - 3 279 Chr. 14 - 100829639 - 100846333 [+] on Build GRCh38
          GenBankmRNA CA949980.1 - 2 134 Chr. 14 - 100832548 - 100860960 [+] on Build GRCh38
          GenBankmRNA AV753036.1 - 430 Chr. - - [-] on Build
          GenBankmRNA N56264.1 - 2 107 Chr. 14 - 100831464 - 100845534 [+] on Build GRCh38
          GenBankmRNA BU079357.1 - 2 113 Chr. 14 - 100836004 - 100861026 [+] on Build GRCh38
          GenBankmRNA BX408226.1 - 3 413 Chr. 14 - 100829491 - 100860979 [+] on Build GRCh38
          GenBankmRNA BQ478473.1 - 2 69 Chr. 14 - 100835852 - 100860906 [+] on Build GRCh38
          GenBankmRNA BI524001.1 - 76 Chr. - - [-] on Build
          GenBankmRNA BX429869.2 - 3 276 Chr. 14 - 100829629 - 100861023 [+] on Build GRCh38
          GenBankmRNA BX367745.1 - 2 512 Chr. 14 - 100835613 - 100860871 [+] on Build GRCh38
          GenBankmRNA BU580862.1 - 3 362 Chr. 14 - 100829558 - 100860756 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0