Search
Search

ADAM8| Gene Symbol: ADAM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109.4 | NP_001100.3 | 9 | 940 | ||
| NM_001164489.1 | NP_001157961.1 | 9 | 940 | |||
| NM_001164490.1 | NP_001157962.1 | 8 | 823 | |||
| XM_011539117.2 | XP_011537419.2 | 10 | 1121 | |||
| XM_017015465.1 | XP_016870954.1 | 10 | 1121 | |||
| XM_017015466.1 | XP_016870955.1 | 10 | 1123 | |||
| GenBank mRNA | AK301147.1 | - | 8 | 788 | ||
| AK304324.1 | - | 9 | 1097 | |||
| AK310662.1 | - | 8 | 796 | |||
| AK312834.1 | - | 9 | 905 | |||
| BC064500.1 | - | 9 | 887 | |||
| BC115404.1 | - | 9 | 859 | 40075505 | ||
| D26579.1 | - | 9 | 863 | |||