Search
Search

Igsf8| Gene Symbol: Igsf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080419.1 | NP_536344.1 | 2 | 312 | ||
| GenBank mRNA | AF411055.1 | - | 2 | 183 | ||
| AF439263.1 | - | 2 | 295 | |||
| AK140254.1 | - | 2 | 325 | |||
| AK146104.1 | - | 2 | 325 | |||
| AK146221.1 | - | 2 | 331 | |||
| AK150239.1 | - | 2 | 331 | |||
| AK158522.1 | - | 2 | 333 | |||
| AK168363.1 | - | 2 | 331 | |||
| AK206992.1 | - | 2 | 164 | |||
| AK213338.1 | - | 2 | 164 | |||
| BC026464.1 | - | 1 | 118 | |||
| BC048387.1 | - | 2 | 312 | 6311441 | ||
| BC060087.1 | - | 940 | ||||