Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PCDH7 SIRNA Assays ›
          Gene Symbol:
          PCDH7
          Gene Name:
          protocadherin 7
          Gene Aliases:
          BH-Pcdh, BHPCDH, PPP1R120
          Chromosome Location:
          Chr.4: 30720378 - 31146801 on Build GRCh38
          UniGene ID:
          Hs.479439
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s10111
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PCDH7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001173523.1 NP_001166994.1 1 3605
          NM_002589.2 NP_002580.2 1 3605
          NM_032456.2 NP_115832.1 1 3612
          NM_032457.3 NP_115833.2 1 3605
          XM_005248163.3 XP_005248220.1 1 3640
          XM_005248164.3 XP_005248221.1 1 3640
          XM_005248166.3 XP_005248223.1 1 3639
          XM_011513842.2 XP_011512144.1 1 3641
          XM_011513843.2 XP_011512145.1 1 3640
          XM_011513844.2 XP_011512146.1 1 3641
          XM_017008259.1 XP_016863748.1 1 3641
          XM_017008260.1 XP_016863749.1 1 3641
          XM_017008261.1 XP_016863750.1 1 3641
          XM_017008262.1 XP_016863751.1 1 3641
          XM_017008263.1 XP_016863752.1 1 3642
          XM_017008264.1 XP_016863753.1 1 3642
          XM_017008265.1 XP_016863754.1 1 3642
          XM_017008266.1 XP_016863755.1 1 3642
          XM_017008267.1 XP_016863756.1 1 3642
          XM_017008268.1 XP_016863757.1 1 3642
          XM_017008269.1 XP_016863758.1 1 3642
          XM_017008270.1 XP_016863759.1 1 3641
          XM_017008271.1 XP_016863760.1 1 3641
          XM_017008272.1 XP_016863761.1 1 3640
          GenBank mRNA AB006755.1 - 1 3607
          AB006756.1 - 1 3164
          AB006757.1 - 2 3311

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001004087, XM_008770189, XM_006251017, XM_006251022, XM_006251023, and others.
          This siRNA also targets the following transcripts in Mouse: NM_018764, NM_001122758, AK165115, BC131967, AK159991, and others.

          Matching Taqman Gene Expression Assay:


          Hs00159908_m1

          Panther Classification:

          Molecular Function -

          cadherin

          Gene Ontology Categories:

          Function(s) Process(es)

          platelet degranulation
          homophilic cell adhesion via plasma membrane adhesion molecules
          calcium ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wrkvw:80/100.66.78.186:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0