Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DICER1 SIRNA Assays ›
Gene Symbol:
Dicer1
Gene Name:
dicer 1, ribonuclease type III
Gene Aliases:
1110006F08Rik, D12Ertd7e, mKIAA0928
Chromosome Location:
Chr.12: 104687740 - 104751952 on Build GRCm38
UniGene ID:
Mm.21135
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s101207
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Dicer1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_148948.2
NP_683750.2
12
2182
GenBank mRNA
AB081470.1
-
11
1980
AF430845.1
-
12
2190
AK129241.1
-
6
741
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008764904.
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
meiotic spindle organization
angiogenesis
regulation of protein phosphorylation
hair follicle development
apoptotic DNA fragmentation
RNA processing
cell proliferation
regulation of Notch signaling pathway
post-embryonic development
zygote asymmetric cell division
regulation of gene expression
negative regulation of Schwann cell proliferation
positive regulation of gene expression
regulation of muscle cell apoptotic process
positive regulation of Schwann cell differentiation
myoblast differentiation involved in skeletal muscle regeneration
rRNA catabolic process
stem cell population maintenance
spinal cord motor neuron differentiation
nerve development
olfactory bulb interneuron differentiation
cerebral cortex development
lung development
embryonic limb morphogenesis
production of siRNA involved in RNA interference
targeting of mRNA for destruction involved in RNA interference
gene silencing by RNA
pre-miRNA processing
hair follicle morphogenesis
regulation of myelination
positive regulation of myelination
peripheral nervous system myelin formation
conversion of ds siRNA to ss siRNA involved in RNA interference
siRNA loading onto RISC involved in RNA interference
embryonic hindlimb morphogenesis
production of miRNAs involved in gene silencing by miRNA
multicellular organism growth
miRNA loading onto RISC involved in gene silencing by miRNA
conversion of ds siRNA to ss siRNA
regulation of odontogenesis of dentin-containing tooth
regulation of viral genome replication
regulation of cell differentiation
regulation of neuron differentiation
positive regulation of transcription from RNA polymerase II promoter
mRNA stabilization
spleen development
digestive tract development
reproductive structure development
regulation of oligodendrocyte differentiation
epidermis morphogenesis
branching morphogenesis of an epithelial tube
neuron projection morphogenesis
anatomical structure development
regulation of neurogenesis
cartilage development
spindle assembly
regulation of RNA metabolic process
defense response to virus
regulation of cell cycle
cardiac muscle cell development
inner ear receptor cell development
negative regulation of glial cell proliferation
intestinal epithelial cell development
cardiac neural crest cell development involved in outflow tract morphogenesis
ganglion development
regulation of enamel mineralization
hair follicle cell proliferation
RNA phosphodiester bond hydrolysis
RNA phosphodiester bond hydrolysis, endonucleolytic
regulation of miRNA metabolic process
positive regulation of miRNA metabolic process
nucleotide binding
nucleic acid binding
RNA binding
double-stranded RNA binding
helicase activity
nuclease activity
endonuclease activity
endoribonuclease activity
ribonuclease III activity
deoxyribonuclease I activity
protein binding
ATP binding
hydrolase activity
endoribonuclease activity, producing 5'-phosphomonoesters
protein domain specific binding
siRNA binding
miRNA binding
metal ion binding
pre-miRNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-fz5f2:80/100.66.72.225:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0