Search
Search

Prepl| Gene Symbol: Prepl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163622.1 | NP_001157094.1 | 4 | 874 | ||
| NM_001163623.1 | NP_001157095.1 | 3 | 338 | |||
| NM_001163624.1 | NP_001157096.1 | 3 | 206 | |||
| NM_145984.3 | NP_666096.3 | 4 | 438 | |||
| GenBank mRNA | AK028877.1 | - | 4 | 832 | ||
| AK050878.1 | - | 3 | 200 | |||
| AK083070.1 | - | 4 | 379 | |||
| AK163108.1 | - | 4 | 839 | |||
| AK195263.1 | - | 1 | 42 | |||
| BC004612.1 | - | 3 | 292 | 3585402 | ||
| BC129880.1 | - | 2 | 870 | |||
| BY257815.1 | - | 3 | 207 | |||