Search
Search

Prepl| Gene Symbol: Prepl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163622.1 | NP_001157094.1 | 10 | 1929 | ||
| NM_001163623.1 | NP_001157095.1 | 9 | 1393 | |||
| NM_001163624.1 | NP_001157096.1 | 9 | 1261 | |||
| NM_145984.3 | NP_666096.3 | 10 | 1493 | |||
| GenBank mRNA | AK028877.1 | - | 10 | 1887 | ||
| AK050878.1 | - | 9 | 1255 | |||
| AK083070.1 | - | 10 | 1434 | |||
| AK129142.1 | - | 4 | 1347 | |||
| BC004612.1 | - | 9 | 1347 | 3585402 | ||