Search
Search

PEPD| Gene Symbol: PEPD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000285.3 | NP_000276.2 | 8 | 721 | ||
| NM_001166057.1 | NP_001159529.1 | 6 | 529 | |||
| GenBank mRNA | AK225037.1 | - | 8 | 602 | ||
| AK290756.1 | - | 8 | 602 | |||
| AK290781.1 | - | 8 | 617 | |||
| AK291561.1 | - | 8 | 619 | |||
| AK293126.1 | - | 6 | 424 | |||
| BC004305.1 | - | 8 | 592 | 3626134 | ||
| BC015027.1 | - | 8 | 622 | 3920275 | ||
| BC028295.1 | - | 8 | 593 | 4470066 | ||
| BT006692.1 | - | 8 | 588 | |||
| CR541669.1 | - | 8 | 588 | |||
| J04605.1 | - | 8 | 604 | |||