Search
Search

Neurl3| Gene Symbol: Neurl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153408.2 | NP_700457.1 | 2 | 151 | ||
| GenBank mRNA | AF321278.1 | - | 2 | 151 | ||
| AK040949.1 | - | 2 | 162 | |||
| AK156207.1 | - | 2 | 162 | |||
| BC056622.1 | - | 2 | 142 | 4989283 | ||