Search
Search

Sec22c| Gene Symbol: Sec22c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164562.1 | NP_001158034.1 | 3 | 464 | ||
| NM_178677.4 | NP_848792.2 | 4 | 589 | |||
| GenBank mRNA | AK029985.1 | - | 4 | 520 | ||
| AK031125.1 | - | 3 | 461 | |||
| AK044249.1 | - | 3 | 464 | |||
| BC046289.1 | - | 3 | 419 | 6469221 | ||