Search
Search

Pomgnt2| Gene Symbol: Pomgnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289558.1 | NP_001276487.1 | 4 | 1604 | ||
| NM_001289559.1 | NP_001276488.1 | 3 | 1504 | |||
| NM_001289560.1 | NP_001276489.1 | 4 | 1600 | |||
| NM_153540.4 | NP_705768.4 | 2 | 1447 | |||
| GenBank mRNA | AK034473.1 | - | 2 | 1433 | ||
| AK042747.1 | - | 4 | 1588 | |||
| AK054403.1 | - | 3 | 1492 | |||
| AK133802.1 | - | 2 | 1430 | |||
| BC025056.1 | - | 2 | 1402 | 4947177 | ||
| BC030931.1 | - | 2 | 1409 | 4914234 | ||