Search
Search

PFKFB1| Gene Symbol: PFKFB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271804.1 | NP_001258733.1 | 8 | 771 | ||
| NM_001271805.1 | NP_001258734.1 | 7 | 672 | |||
| NM_002625.3 | NP_002616.2 | 8 | 837 | |||
| NR_073450.1 | 7 | 743 | ||||
| XM_017029576.1 | XP_016885065.1 | 8 | 1241 | |||
| XM_017029577.1 | XP_016885066.1 | 8 | 1241 | |||
| XM_017029578.1 | XP_016885067.1 | 9 | 1323 | |||
| GenBank mRNA | AK300724.1 | - | 7 | 672 | ||
| AK314089.1 | - | 8 | 1053 | |||
| BC096078.2 | - | 8 | 839 | 40000711 | ||
| BC096079.3 | - | 8 | 838 | 40000713 | ||
| JQ425694.1 | - | 7 | 687 | |||
| JQ425695.1 | - | 8 | 715 | |||
| M19938.1 | - | 6 | 487 | |||
| X52638.1 | - | 8 | 845 | |||