Search
Search

Cep68| Gene Symbol: Cep68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172260.3 | NP_758464.2 | 3 | 1114 | ||
| GenBank mRNA | AK031622.1 | - | 3 | 1111 | ||
| AK140820.1 | - | 3 | 1115 | |||
| AK145286.1 | - | 3 | 1083 | |||
| AK146669.1 | - | 3 | 1062 | |||
| AK148267.1 | - | 1 | 254 | |||
| AK162594.1 | - | 3 | 1120 | |||
| AK169934.1 | - | 3 | 1110 | |||
| AK172975.1 | - | 3 | 1045 | |||
| BC027174.1 | - | 3 | 1108 | 3154964 | ||