Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GSN SIRNA Assays ›
          Gene Symbol:
          Gsn
          Gene Name:
          gelsolin
          Gene Aliases:
          ADF
          Chromosome Location:
          Chr.2: 35256359 - 35307902 on Build GRCm38
          UniGene ID:
          Mm.21109
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s105803

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gsn
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001206367.1 NP_001193296.1 6 758
          NM_001206368.1 NP_001193297.1 5 674
          NM_001206369.1 NP_001193298.1 5 719
          NM_146120.4 NP_666232.2 5 781
          GenBank mRNA AK076156.1 - 5 721
          AK089934.1 - 6 713
          AK135632.1 - 4 494
          AK143664.1 - 5 748
          AK146969.1 - 6 675
          AK148957.1 - 6 738
          AK150359.1 - 6 681
          AK151684.1 - 6 700
          AK153029.1 - 6 678
          AK166831.1 - 5 694
          AK166932.1 - 6 672
          AK167918.1 - 6 688
          AK168461.1 - 6 691
          AK168532.1 - 6 692
          AK168707.1 - 5 627
          AK168725.1 - 6 675
          AK168829.1 - 5 726
          AK169041.1 - 6 678
          AK169670.1 - 6 703
          AK196436.1 - 2 170
          BC023143.1 - 5 719 3711602
          BC060377.1 - 6 706 4983664
          J04953.1 - 4 568

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          phagocytosis, engulfment
          apoptotic process
          actin polymerization or depolymerization
          positive regulation of gene expression
          striated muscle atrophy
          vesicle-mediated transport
          cell projection organization
          actin filament polymerization
          regulation of cell adhesion
          protein destabilization
          sequestering of actin monomers
          actin nucleation
          negative regulation of viral entry into host cell
          phosphatidylinositol-mediated signaling
          actin filament severing
          barbed-end actin filament capping
          positive regulation of actin nucleation
          actin filament capping
          cilium morphogenesis
          cellular response to cadmium ion
          regulation of podosome assembly
          actin filament reorganization
          renal protein absorption
          hepatocyte apoptotic process
          positive regulation of keratinocyte apoptotic process
          regulation of wound healing, spreading of epidermal cells
          regulation of establishment of T cell polarity
          regulation of plasma membrane raft polarization
          regulation of receptor clustering
          positive regulation of protein processing in phagocytic vesicle
          amyloid fibril formation
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          actin binding
          calcium ion binding
          protein binding
          protein domain specific binding
          myosin II binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0