Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PLAU SIRNA Assays ›
          Gene Symbol:
          PLAU
          Gene Name:
          plasminogen activator, urokinase
          Gene Aliases:
          ATF, BDPLT5, QPD, UPA, URK, u-PA
          Chromosome Location:
          Chr.10: 73909182 - 73917501 on Build GRCh38
          UniGene ID:
          Hs.77274
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s10610

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PLAU
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145031.2 NP_001138503.1 8, 9 1413
          NM_001319191.1 NP_001306120.1 8, 9 1100
          NM_002658.4 NP_002649.1 9, 10 1128
          XM_011539866.2 XP_011538168.1 9, 10 1123
          GenBank mRNA A18397.1 - 9, 10 1078
          A35395.1 - 9, 10 1048
          AB208961.1 - 3, 4 477
          AK297940.1 - 8, 9 1069
          AK298560.1 - 8, 9 1382
          AK312960.1 - 9, 10 1097
          AY820134.1 - 7, 8 919
          BC013575.2 - 9, 10 1071 3890980
          BT007391.1 - 8, 9 979
          D00244.1 - 9, 10 1045
          D11143.1 - 7, 8 919
          M15476.1 - 9, 10 1055
          X02760.1 - 8, 9 1019
          Z36790.1 - 1, 2 67

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          proteolysis
          chemotaxis
          signal transduction
          spermatogenesis
          embryo implantation
          blood coagulation
          positive regulation of cell proliferation
          regulation of receptor activity
          response to activity
          smooth muscle cell migration
          regulation of smooth muscle cell migration
          positive regulation of smooth muscle cell migration
          positive regulation of cell migration
          regulation of cell adhesion mediated by integrin
          cellular response to hepatocyte growth factor stimulus
          fibrinolysis
          skeletal muscle tissue regeneration
          response to hyperoxia
          positive regulation of ovulation
          regulation of wound healing
          neuron death
          cellular response to lipopolysaccharide
          cellular response to glucose stimulus
          cellular response to hypoxia
          cellular response to fluid shear stress
          cellular response to staurosporine
          regulation of smooth muscle cell-matrix adhesion
          regulation of hepatocyte proliferation
          positive regulation of reactive oxygen species metabolic process
          serine-type endopeptidase activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0