Search
Search

Nudt6| Gene Symbol: Nudt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291044.1 | NP_001277973.1 | 5 | 853 | ||
| NM_153561.3 | NP_705789.1 | 5 | 850 | |||
| GenBank mRNA | AF453427.1 | - | 5 | 962 | ||
| AF453428.1 | - | 5 | 839 | |||
| AK145299.1 | - | 5 | 845 | |||
| BC027267.1 | - | 5 | 844 | 3500531 | ||
| BC146000.1 | - | 5 | 847 | 40131303 | ||