Search
Search

Map7d1| Gene Symbol: Map7d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145970.1 | NP_001139442.1 | 5 | 911 | ||
| NM_144941.3 | NP_659190.3 | 5 | 911 | |||
| GenBank mRNA | AK151613.1 | - | 5 | 899 | ||
| BC023677.1 | - | 5 | 886 | 5354497 | ||
| BC023774.1 | - | 5 | 908 | |||
| BM944941.1 | - | 3 | 202 | |||
| BQ713545.1 | - | 4 | 280 | |||
| CV563343.1 | - | 4 | 482 | |||