Search
Search

Usp7| Gene Symbol: Usp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003918.2 | NP_001003918.2 | 11 | 1190 | ||
| GenBank mRNA | AF548565.1 | - | 11 | 1133 | ||
| AK135814.1 | - | 1382 | ||||
| BC046963.1 | - | 11 | 1128 | |||
| BC100666.1 | - | 6 | 496 | |||