Search
Search

| Gene Symbol: Olfr203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146486.2 | NP_666697.2 | 1 | 245 | ||
| GenBank mRNA | BC128025.1 | - | 1 | 248 | 40136560 | |
| Gene Symbol: Olfr194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005524.2 | NP_001005524.2 | 1 | 245 | ||