Search
Search

PSG9| Gene Symbol: PSG9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301707.1 | NP_001288636.1 | 2 | 368 | ||
| NM_001301708.1 | NP_001288637.1 | 2 | 368 | |||
| NM_001301709.1 | NP_001288638.1 | 2 | 368 | |||
| NM_002784.4 | NP_002775.3 | 2 | 368 | |||
| XM_005259075.3 | XP_005259132.1 | 2 | 330 | |||
| XM_017027004.1 | XP_016882493.1 | 2 | 330 | |||
| XM_017027005.1 | XP_016882494.1 | 2 | 363 | |||
| XM_017027006.1 | XP_016882495.1 | 2 | 363 | |||
| XM_017027007.1 | XP_016882496.1 | 2 | 330 | |||
| GenBank mRNA | AK300187.1 | - | 3 | 214 | ||
| AK300229.1 | - | 2 | 333 | |||
| AK313256.1 | - | 2 | 335 | |||
| BC005925.1 | - | 2 | 332 | 4044135 | ||
| BC020759.1 | - | 2 | 333 | 4731739 | ||
| BX952516.1 | - | 2 | 336 | |||
| CR407605.1 | - | 2 | 236 | |||
| DC380417.1 | - | 2 | 368 | |||
| M34421.1 | - | 2 | 314 | |||
| M34481.1 | - | 2 | 305 | |||
| M38046.1 | - | 1 | 94 | |||
| M58591.1 | - | 2 | 307 | |||
| M94890.1 | - | 2 | 303 | |||
| U25987.1 | - | 2 | 246 | |||
| X17610.1 | - | 2 | 314 | |||