Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTAFR SIRNA Assays ›
          Gene Symbol:
          PTAFR
          Gene Name:
          platelet activating factor receptor
          Gene Aliases:
          PAFR
          Chromosome Location:
          Chr.1: 28147166 - 28193936 on Build GRCh38
          UniGene ID:
          Hs.77542
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s11433

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTAFR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000952.4 NP_000943.1 2 1277
          NM_001164721.1 NP_001158193.1 3 1221
          NM_001164722.2 NP_001158194.1 2 1139
          NM_001164723.2 NP_001158195.1 3 1348
          GenBank mRNA AK290240.1 - 2 1023
          AK292053.1 - 3 1221
          AK313174.1 - 2 1008
          AY275466.1 - 1 886
          BC013816.2 - 1 926 4343214
          BC063000.1 - 2 1013 5176867
          BT009801.1 - 1 886
          CD013879.1 - 3 1077
          D10202.1 - 2 998
          L07334.1 - 1 886
          M76674.1 - 1 911
          M80436.1 - 2 1152

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokine production
          regulation of transcription from RNA polymerase II promoter
          chemotaxis
          inflammatory response
          immune response
          G-protein coupled receptor signaling pathway
          parturition
          response to symbiotic bacterium
          positive regulation of phospholipase C activity
          lipopolysaccharide-mediated signaling pathway
          positive regulation of tumor necrosis factor production
          inositol trisphosphate biosynthetic process
          G-protein coupled purinergic nucleotide receptor signaling pathway
          positive regulation of neutrophil degranulation
          transcytosis
          positive regulation of interleukin-6 biosynthetic process
          positive regulation of translation
          negative regulation of blood pressure
          positive regulation of vasoconstriction
          positive regulation of vasodilation
          phosphatidylinositol-mediated signaling
          positive regulation of smooth muscle cell proliferation
          interferon-gamma-mediated signaling pathway
          positive regulation of inositol phosphate biosynthetic process
          cellular response to gravity
          cellular response to cAMP
          cellular response to fatty acid
          response to dexamethasone
          positive regulation of voltage-gated chloride channel activity
          positive regulation of leukocyte tethering or rolling
          positive regulation of sensory perception of pain
          positive regulation of transcytosis
          positive regulation of maternal process involved in parturition
          positive regulation of gastro-intestinal system smooth muscle contraction
          cellular response to 2-O-acetyl-1-O-hexadecyl-sn-glycero-3-phosphocholine
          lipopolysaccharide binding
          lipopolysaccharide receptor activity
          G-protein coupled receptor activity
          platelet activating factor receptor activity
          phospholipid binding
          G-protein coupled purinergic nucleotide receptor activity
          mitogen-activated protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0