Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTN SIRNA Assays ›
          Gene Symbol:
          PTN
          Gene Name:
          pleiotrophin
          Gene Aliases:
          HARP, HB-GAM, HBBM, HBGF-8, HBGF8, HBNF, HBNF-1, NEGF1, OSF-1
          Chromosome Location:
          Chr.7: 137227341 - 137343865 on Build GRCh38
          UniGene ID:
          Hs.371249
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s11502

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321386.1 NP_001308315.1 4, 5 579
          NM_002825.5 NP_002816.1 4, 5 816
          GenBank mRNA AK290488.1 - 4, 5 645
          AK313424.1 - 4, 5 719
          BC005916.1 - 4, 5 804 4248708
          BI462078.1 - 4, 5 588
          BT019692.1 - 3, 4 453
          CN413530.1 - 4, 5 565
          CR450338.1 - 3, 4 453
          D90226.1 - 4, 5 577
          M57399.1 - 4, 5 848
          X52946.1 - 4, 5 704

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          transmembrane receptor protein tyrosine phosphatase signaling pathway
          nervous system development
          heart development
          learning
          positive regulation of cell proliferation
          regulation of cell shape
          positive regulation of cell-substrate adhesion
          positive regulation of neuron projection development
          response to activity
          negative regulation of angiogenesis
          spinal cord development
          cerebellum development
          thalamus development
          bone mineralization
          lung development
          negative regulation of cell migration
          response to estradiol
          response to progesterone
          cellular response to UV
          cellular response to platelet-derived growth factor stimulus
          response to drug
          positive regulation of apoptotic process
          negative regulation of catalytic activity
          estrous cycle
          endothelial cell differentiation
          negative regulation of membrane potential
          negative regulation of epithelial cell proliferation
          positive regulation of cell division
          retinal rod cell differentiation
          negative regulation of glial cell proliferation
          long-term synaptic potentiation
          cellular response to vitamin D
          cellular response to hypoxia
          negative regulation of mesenchymal cell proliferation
          response to kainic acid
          rod bipolar cell differentiation
          response to ciliary neurotrophic factor
          positive regulation of skeletal muscle acetylcholine-gated channel clustering
          negative regulation of neuromuscular junction development
          response to nerve growth factor
          positive regulation of hepatocyte proliferation
          protein phosphatase inhibitor activity
          growth factor activity
          heparin binding
          chondroitin sulfate proteoglycan binding
          chondroitin sulfate binding
          vascular endothelial growth factor binding
          heparan sulfate binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline