Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB27B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004163.4 | NP_004154.2 | 6 | 786 | ||
XM_005266733.1 | XP_005266790.1 | 7 | 707 | |||
XM_017025913.1 | XP_016881402.1 | 9 | 1540 | |||
XM_017025914.1 | XP_016881403.1 | 8 | 935 | |||
GenBank mRNA | AF329499.1 | - | 6 | 786 | ||
AF498954.1 | - | 5 | 543 | |||
AK314115.1 | - | 6 | 1064 | |||
BC027474.1 | - | 6 | 654 | 4520182 | ||
U57093.2 | - | 6 | 635 |