Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RAD51C SIRNA Assays ›
          Gene Symbol:
          RAD51C
          Gene Name:
          RAD51 paralog C
          Gene Aliases:
          BROVCA3, FANCO, R51H3, RAD51L2
          Chromosome Location:
          Chr.17: 58692538 - 58734342 on Build GRCh38
          UniGene ID:
          Hs.412587
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s11738

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RAD51C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002876.3 NP_002867.1 2 298
          NM_058216.2 NP_478123.1 2 298
          NR_103872.1 2 298
          NR_103873.1 2 195
          XM_006722001.3 XP_006722064.1 2 269
          XM_006722002.3 XP_006722065.1 2 269
          XM_006722004.3 XP_006722067.1 2 191
          XM_006722005.3 XP_006722068.1 2 177
          XM_011525092.2 XP_011523394.1 2 397
          XM_011525093.2 XP_011523395.1 2 558
          XM_011525094.2 XP_011523396.1 2 246
          XM_017024914.1 XP_016880403.1 2 191
          XM_017024915.1 XP_016880404.1 2 543
          XM_017024916.1 XP_016880405.1 3 451
          XM_017024917.1 XP_016880406.1 2 176
          XM_017024918.1 XP_016880407.1 2 245
          XM_017024919.1 XP_016880408.1 2 558
          GenBank mRNA AA304618.1 - 2 129
          AF029669.1 - 2 269
          AF029670.1 - 2 243
          AK290487.1 - 2 269
          AK303363.1 - 2 208
          BC000667.2 - 2 256
          BC093930.1 - 2 264
          BC101485.1 - 2 264
          BC107753.1 - 2 302 6715697
          BC143259.1 - 2 161
          BT007339.1 - 2 227

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance via recombination
          double-strand break repair via homologous recombination
          DNA recombinase assembly
          DNA synthesis involved in DNA repair
          strand displacement
          DNA repair
          DNA recombination
          mitotic recombination
          sister chromatid cohesion
          female meiosis sister chromatid cohesion
          reciprocal meiotic recombination
          male meiosis I
          spermatogenesis
          blood coagulation
          response to ionizing radiation
          positive regulation of G2/M transition of mitotic cell cycle
          strand invasion
          chromosome organization involved in meiotic cell cycle
          recombinase activity
          four-way junction DNA binding
          DNA binding
          double-stranded DNA binding
          single-stranded DNA binding
          protein binding
          ATP binding
          DNA-dependent ATPase activity
          crossover junction endodeoxyribonuclease activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0