Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SSXB2 SIRNA Assays ›
          Gene Symbol:
          Ssxb2 Gm14459 Gm6592 Ssxb1 Ssxb10 Ssxb11 Ssxb12 Ssxb3 Ssxb5 Ssxb6 Ssxb8 Ssxb9
          Gene Name:
          synovial sarcoma, X member B, breakpoint 2
          predicted gene 14459
          predicted gene 6592
          synovial sarcoma, X member B, breakpoint 1
          synovial sarcoma, X member B, breakpoint 10
          synovial sarcoma, X member B, breakpoint 11
          synovial sarcoma, X member B, breakpoint 12
          synovial sarcoma, X member B, breakpoint 3
          synovial sarcoma, X member B, breakpoint 5
          synovial sarcoma, X member B, breakpoint 6
          synovial sarcoma, X member B, breakpoint 8
          synovial sarcoma, X member B, breakpoint 9
          Gene Aliases:
          -
          OTTMUSG00000016698
          EG625480
          4930414C09Rik
          -
          -
          -
          -
          -
          -
          -
          -
          Chromosome Location:
          Chr.X: 8454343 - 8461726 on Build GRCm38
          Chromosome Location:
          Chr.X: 8513543 - 8526726 on Build GRCm38
          Chromosome Location:
          Chr.X: 8843994 - 8849607 on Build GRCm38
          Chromosome Location:
          Chr.X: 8413305 - 8422157 on Build GRCm38
          Chromosome Location:
          Chr.X: 8327424 - 8336235 on Build GRCm38
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          Chr.X: 8583404 - 8591810 on Build GRCm38
          Chromosome Location:
          Chr.X: 8803691 - 8809445 on Build GRCm38
          Chromosome Location:
          Chr.X: 8542179 - 8548362 on Build GRCm38
          Chromosome Location:
          Chr.X: 8684843 - 8690883 on Build GRCm38
          Chromosome Location:
          Chr.X: 8366978 - 8375388 on Build GRCm38
          UniGene ID:
          Mm.379565
          Mm.475615
          Mm.430078
          Mm.298030
          Mm.484393
          Mm.298030
          Mm.298030
          Mm.379565
          Mm.227643
          Mm.227643
          Mm.379565
          Mm.379565
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s118674

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ssxb2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001450.4 NP_001001450.1 6 689
          NM_001134226.1 NP_001127698.1 5 512
          GenBank mRNA AK076879.1 - 6 690
          AY244793.1 - 6 690
          BC048441.1 - 5 512 6773922
          BC089478.1 - 5 512 6743842
          Gene Symbol: Gm14459
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001126491.1 NP_001119963.1 6 484
          Gene Symbol: Gm6592
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001081564.1 NP_001075033.1 6 451
          Gene Symbol: Ssxb1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_026492.3 NP_080768.1 7 638
          GenBank mRNA AK006218.1 - 7 639
          AK015135.1 - 7 639
          AY244792.1 - 7 639
          BC145538.1 - 7 611 9054101
          BC145541.1 - 556 9054104
          BC145782.1 - 7 530 40131085
          BC145784.1 - 7 530 40131087
          BC171964.1 - 611 9054250
          Gene Symbol: Ssxb10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_199064.1 NP_951019.1 7 639
          GenBank mRNA AY244801.1 - 7 639
          Gene Symbol: Ssxb11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AY244802.1 - 6 502
          Gene Symbol: Ssxb12
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AY244803.1 - 6 502
          Gene Symbol: Ssxb3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_198898.2 NP_942601.1 6 503
          GenBank mRNA AY244794.1 - 6 503
          BC087939.1 - 7 583
          BC132618.1 - 6 503 40130895
          BC132644.1 - 6 503 40130921
          Gene Symbol: Ssxb5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_199319.3 NP_955023.1 6 502
          GenBank mRNA AY244796.1 - 6 502
          Gene Symbol: Ssxb6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001205108.1 NP_001192037.1 462
          Gene Symbol: Ssxb8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001081565.3 NP_001075034.3 6 462
          Gene Symbol: Ssxb9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_199066.2 NP_951021.2 7 639
          GenBank mRNA AY244800.1 - 639

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0