Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AHCY SIRNA Assays ›
          Gene Symbol:
          AHCY
          Gene Name:
          adenosylhomocysteinase
          Gene Aliases:
          SAHH, adoHcyase
          Chromosome Location:
          Chr.20: 34235012 - 34311976 on Build GRCh38
          UniGene ID:
          Hs.388004
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1197
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AHCY
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000687.3 NP_000678.1 5, 6 809
          NM_001161766.1 NP_001155238.1 5, 6 871
          NM_001322084.1 NP_001309013.1 5, 6 1789
          NM_001322085.1 NP_001309014.1 5, 6 1611
          NM_001322086.1 NP_001309015.1 5, 6 1800
          XM_005260317.2 XP_005260374.1 5, 6 1104
          XM_011528656.2 XP_011526958.2 5, 6 678
          XM_011528657.2 XP_011526959.2 5, 6 679
          XM_011528658.2 XP_011526960.2 5, 6 679
          XM_011528659.1 XP_011526961.1 5, 6 699
          XM_017027708.1 XP_016883197.1 5, 6 657
          XM_017027709.1 XP_016883198.1 5, 6 657
          XM_017027710.1 XP_016883199.1 5, 6 708
          GenBank mRNA AK097610.1 - 5, 6 860
          AK290422.1 - 5, 6 653
          AK300014.1 - 5, 6 590
          AK316180.1 - 5, 6 563
          BC010018.2 - 5, 6 637 2905378
          BC011606.2 - 5, 6 642 3139231
          BC114924.1 - 3, 4 357 40025256
          BT006697.1 - 5, 6 568
          M61831.1 - 5, 6 615
          M61832.1 - 5, 6 611

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_017201, M15185, BC061841.
          This siRNA also targets the following transcripts in Mouse: NM_016661, AK075629, AK153421, AK213514, AK183971, and others.

          Matching Taqman Gene Expression Assay:


          Hs00426322_m1

          Panther Classification:

          Molecular Function -

          hydrolase

          Gene Ontology Categories:

          Function(s) Process(es)

          sulfur amino acid metabolic process
          response to hypoxia
          chronic inflammatory response to antigenic stimulus
          one-carbon metabolic process
          response to nutrient
          S-adenosylhomocysteine catabolic process
          methylation
          S-adenosylmethionine cycle
          circadian sleep/wake cycle
          homocysteine biosynthetic process
          adenosylhomocysteinase activity
          protein binding
          adenyl nucleotide binding
          identical protein binding
          NAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0