Search
Search

Gm16445
4921511M17Rik
Gm10057
Gm15140
Samt1| Gene Symbol: Gm16445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243032.1 | NP_001229961.1 | 3 | 366 | ||
| Gene Symbol: 4921511M17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201358.1 | NP_001188287.1 | 377 | |||
| Gene Symbol: Gm10057 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243016.1 | NP_001229945.1 | 379 | |||
| Gene Symbol: Gm15140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243017.1 | NP_001229946.1 | 377 | |||
| Gene Symbol: Samt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030036.1 | NP_084312.1 | 377 | |||
| GenBank mRNA | AK019538.1 | - | 378 | |||