Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HSPA8 SIRNA Assays ›
Gene Symbol:
Hspa8
Gene Name:
heat shock protein 8
Gene Aliases:
2410008N15Rik, Hsc70, Hsc71, Hsc73, Hsp73, Hspa10
Chromosome Location:
Chr.9: 40801273 - 40805199 on Build GRCm38
UniGene ID:
Mm.290774
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s125650
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hspa8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031165.4
NP_112442.2
3
394
GenBank mRNA
AK004608.1
-
2
683
AK035286.1
-
3
392
AK075935.1
-
3
391
AK081701.1
-
3
392
AK098074.1
-
3
391
AK145416.1
-
3
314
AK145579.1
-
3
388
AK146547.1
-
3
395
AK146708.1
-
3
389
AK146985.1
-
3
392
AK147864.1
-
3
392
AK149954.1
-
3
389
AK150474.1
-
3
386
AK150498.1
-
3
386
AK150701.1
-
3
396
AK150958.1
-
3
389
AK151041.1
-
3
392
AK151065.1
-
3
392
AK151127.1
-
3
395
AK151287.1
-
3
392
AK151435.1
-
3
392
AK151516.1
-
3
398
AK151537.1
-
3
389
AK151741.1
-
3
392
AK151775.1
-
3
394
AK151808.1
-
3
395
AK151865.1
-
3
392
AK151892.1
-
3
394
AK151948.1
-
3
395
AK151997.1
-
3
392
AK152315.1
-
3
395
AK152598.1
-
3
392
AK152697.1
-
3
392
AK152703.1
-
3
392
AK152768.1
-
3
395
AK152803.1
-
3
395
AK153032.1
-
3
394
AK153834.1
-
3
392
AK157831.1
-
3
392
AK159479.1
-
3
392
AK162775.1
-
3
392
AK163987.1
-
3
392
AK164000.1
-
3
392
AK166545.1
-
3
394
AK166643.1
-
3
406
AK166721.1
-
3
389
AK166767.1
-
3
392
AK166776.1
-
3
395
AK166808.1
-
3
392
AK166830.1
-
3
390
AK166846.1
-
3
392
AK166861.1
-
3
395
AK166873.1
-
3
392
AK166908.1
-
3
395
AK166910.1
-
3
392
AK166913.1
-
3
392
AK166933.1
-
3
392
AK167043.1
-
3
394
AK167121.1
-
3
395
AK167122.1
-
3
392
AK167134.1
-
3
390
AK167163.1
-
3
394
AK167218.1
-
3
392
AK167229.1
-
3
393
AK167254.1
-
3
393
AK167414.1
-
3
392
AK167845.1
-
3
395
AK167910.1
-
3
395
AK168435.1
-
3
395
AK168492.1
-
3
394
AK168519.1
-
3
392
AK168542.1
-
3
394
AK168577.1
-
3
392
AK168625.1
-
3
392
AK168654.1
-
3
392
AK168711.1
-
3
392
AK168750.1
-
3
395
AK168776.1
-
3
395
AK168887.1
-
3
394
AK168934.1
-
3
395
AK169093.1
-
3
392
AK169179.1
-
3
392
AK169236.1
-
3
392
AK169293.1
-
3
406
AK169329.1
-
3
395
AK169594.1
-
3
391
AK171434.1
-
3
393
AK185576.1
-
2
153
AK188190.1
-
1
100
AK188441.1
-
1
100
AK190603.1
-
1
100
AK199049.1
-
1
100
AK204952.1
-
1
100
AK206788.1
-
1
100
AK212540.1
-
1
100
AK218563.1
-
1
100
AK220016.1
-
1
100
BC006722.1
-
3
416
3711399
BC066191.1
-
3
348
30467052
BC085486.1
-
3
380
30540914
BC089322.1
-
3
373
30538640
BC089457.1
-
3
351
30628505
BC094900.1
-
3
365
30063483
BC106169.1
-
1
72
BC106193.1
-
3
376
6329283
U27129.1
-
3
343
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
protein folding
RNA splicing
protein refolding
negative regulation of transcription, DNA-templated
ATP metabolic process
positive regulation of mRNA splicing, via spliceosome
chaperone mediated protein folding requiring cofactor
regulation of cell cycle
chaperone-mediated protein folding
clathrin coat disassembly
negative regulation of fibril organization
nucleotide binding
G-protein coupled receptor binding
receptor binding
protein binding
ATP binding
ATPase activity
enzyme binding
MHC class II protein complex binding
heat shock protein binding
ubiquitin protein ligase binding
ATPase activity, coupled
ADP binding
poly(A) RNA binding
unfolded protein binding
C3HC4-type RING finger domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-29f27:80/100.66.78.233:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0