Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCL3 SIRNA Assays ›
          Gene Symbol:
          CCL3 CCL3L1 CCL3L3
          Gene Name:
          C-C motif chemokine ligand 3
          C-C motif chemokine ligand 3 like 1
          C-C motif chemokine ligand 3 like 3
          Gene Aliases:
          G0S19-1, LD78ALPHA, MIP-1-alpha, MIP1A, SCYA3
          464.2, D17S1718, G0S19-2, LD78, LD78-beta(1-70), LD78BETA, MIP1AP, SCYA3L, SCYA3L1
          464.2, D17S1718, G0S19-2, LD78, LD78BETA, SCYA3L, SCYA3L1
          Chromosome Location:
          Chr.17: 36088256 - 36090160 on Build GRCh38
          Chromosome Location:
          -
          Chromosome Location:
          Chr.17: 36194869 - 36196748 on Build GRCh38
          UniGene ID:
          Hs.514107
          Hs.512683
          Hs.454713
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID s12570

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CCL3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002983.2 NP_002974.1 3 528
          GenBank mRNA BC071834.1 - 3 500 5935287
          BC171831.1 - 3 462 9054485
          BC171891.1 - 3 462 9054545
          BX103455.1 - 3 528
          D00044.1 - 3 509
          M23452.1 - 3 509
          M25315.1 - 3 509
          X03754.1 - 3 509
          Gene Symbol: CCL3L1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021006.5 NP_066286.1 3 514
          NR_111964.1 3 706
          GenBank mRNA BC027888.1 - 3 502 5222247
          BC107710.1 - 3 518 6699133
          BC146914.1 - 3 465 9056842
          X52149.1 - 3 692
          Gene Symbol: CCL3L3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001437.3 NP_001001437.2 3 504
          GenBank mRNA BC007783.2 - 3 499 4308472
          BC146919.1 - 3 465 9056847
          BC147008.1 - 3 465 9056936
          BC147013.1 - 3 465 9056941
          CF128666.1 - 3 483

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cytokine intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          osteoblast differentiation
          cell activation
          monocyte chemotaxis
          calcium ion transport
          cellular calcium ion homeostasis
          exocytosis
          chemotaxis
          inflammatory response
          cytoskeleton organization
          G-protein coupled receptor signaling pathway
          cell-cell signaling
          behavior
          regulation of cell shape
          response to toxic substance
          positive regulation of gene expression
          negative regulation of gene expression
          T cell chemotaxis
          release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          calcium-mediated signaling
          signaling
          positive regulation of cell migration
          negative regulation of bone mineralization
          neutrophil chemotaxis
          lipopolysaccharide-mediated signaling pathway
          positive regulation of tumor necrosis factor production
          eosinophil degranulation
          protein kinase B signaling
          positive regulation of neuron apoptotic process
          positive regulation of GTPase activity
          astrocyte cell migration
          negative regulation by host of viral transcription
          negative regulation of osteoclast differentiation
          eosinophil chemotaxis
          macrophage chemotaxis
          lymphocyte chemotaxis
          positive regulation of interleukin-1 beta secretion
          positive regulation of inflammatory response
          positive regulation of calcium-mediated signaling
          positive chemotaxis
          positive regulation of protein kinase B signaling
          positive regulation of calcium ion transport
          regulation of sensory perception of pain
          chemokine-mediated signaling pathway
          positive regulation of ERK1 and ERK2 cascade
          response to cholesterol
          cellular response to interferon-gamma
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          cellular response to organic cyclic compound
          granulocyte chemotaxis
          positive regulation of calcium ion import
          positive regulation of natural killer cell chemotaxis
          negative regulation of cell proliferation
          protein kinase activity
          calcium-dependent protein kinase C activity
          protein binding
          chemokine activity
          phospholipase activator activity
          kinase activity
          CCR1 chemokine receptor binding
          CCR5 chemokine receptor binding
          chemoattractant activity
          identical protein binding
          CCR chemokine receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0