Search
Search

CCL13| Gene Symbol: CCL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005408.2 | NP_005399.1 | 2 | 192 | ||
| GenBank mRNA | AJ001634.1 | - | 2 | 159 | ||
| BC008621.1 | - | 2 | 155 | 4184250 | ||
| BT007385.1 | - | 2 | 117 | |||
| CR450337.1 | - | 2 | 117 | |||
| U46767.1 | - | 2 | 150 | |||
| U59808.1 | - | 2 | 192 | |||
| X98306.1 | - | 2 | 146 | |||
| Z77650.1 | - | 2 | 69 | |||
| Z77651.1 | - | 1 | 31 | |||