Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SDCBP SIRNA Assays ›
          Gene Symbol:
          SDCBP SDCBPP2
          Gene Name:
          syndecan binding protein
          syndecan binding protein pseudogene 2
          Gene Aliases:
          MDA-9, MDA9, ST1, SYCL, TACIP18
          -
          Chromosome Location:
          Chr.8: 58553169 - 58582860 on Build GRCh38
          Chromosome Location:
          Chr.8: 69942921 - 69944286 on Build GRCh38
          UniGene ID:
          Hs.200804
          Hs.454036
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s12643

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SDCBP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001007067.1 NP_001007068.1 6 702
          NM_001007068.1 NP_001007069.1 6 687
          NM_001007069.1 NP_001007070.1 6 702
          NM_001007070.1 NP_001007071.1 6 699
          NM_005625.3 NP_005616.2 6 705
          XM_011517571.2 XP_011515873.2 6 2074
          XM_017013727.1 XP_016869216.1 6 2077
          XM_017013728.1 XP_016869217.1 6 2074
          GenBank mRNA AF000652.1 - 6 703
          AF006636.1 - 6 630
          AK128645.1 - 7 2129
          AK295198.1 - 5 442
          AK298697.1 - 6 517
          AK300647.1 - 6 619
          AK312274.1 - 6 613
          BC013254.2 - 6 589
          BC113674.1 - 6 649 8322726
          BC113676.1 - 6 649 8322728
          BC143915.1 - 6 646 9052433
          BC143916.1 - 6 649 9052434
          U83463.1 - 5 579
          Gene Symbol: SDCBPP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC030617.1 - 1 910
          BC045716.1 - 1 910

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_031986, AJ292243, AF248548, BC064651.

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of receptor internalization
          protein targeting to membrane
          substrate-dependent cell migration, cell extension
          Ras protein signal transduction
          chemical synaptic transmission
          regulation of mitotic cell cycle
          positive regulation of cell proliferation
          positive regulation of epithelial to mesenchymal transition
          positive regulation of pathway-restricted SMAD protein phosphorylation
          actin cytoskeleton organization
          positive regulation of cell growth
          positive regulation of cell migration
          positive regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of proteasomal ubiquitin-dependent protein catabolic process
          intracellular signal transduction
          positive regulation of phosphorylation
          positive regulation of JNK cascade
          ephrin receptor signaling pathway
          cell-cell adhesion
          positive regulation of exosomal secretion
          positive regulation of extracellular exosome assembly
          glycoprotein binding
          frizzled binding
          interleukin-5 receptor binding
          protein binding
          protein C-terminus binding
          cytoskeletal adaptor activity
          growth factor binding
          neurexin family protein binding
          identical protein binding
          protein homodimerization activity
          syndecan binding
          ephrin receptor binding
          protein heterodimerization activity
          protein N-terminus binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-n8jtk:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0