Search
Search

Gstm2| Gene Symbol: Gstm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177426.1 | NP_803175.1 | 476 | |||
| GenBank mRNA | BC158576.1 | - | 5 | 1685 | ||
| FJ179402.1 | - | 476 | ||||
| FQ214985.1 | - | 7 | 509 | |||
| FQ218547.1 | - | 7 | 542 | |||
| FQ218596.1 | - | 7 | 509 | |||
| FQ219549.1 | - | 7 | 511 | |||
| FQ219560.1 | - | 7 | 509 | |||
| J02592.1 | - | 5 | 380 | |||
| M13590.1 | - | 6 | 406 | |||