Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SHC1 SIRNA Assays ›
          Gene Symbol:
          SHC1
          Gene Name:
          SHC adaptor protein 1
          Gene Aliases:
          SHC, SHCA
          Chromosome Location:
          Chr.1: 154962298 - 154974492 on Build GRCh38
          UniGene ID:
          Hs.433795
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s12812

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SHC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130040.1 NP_001123512.1 7 1154
          NM_001130041.1 NP_001123513.1 8 842
          NM_001202859.1 NP_001189788.1 8 842
          NM_003029.4 NP_003020.2 8 842
          NM_183001.4 NP_892113.4 7 1154
          XM_005245449.4 XP_005245506.1 7 2370
          XM_005245451.4 XP_005245508.1 8 765
          XM_011509892.2 XP_011508194.1 7 2367
          XM_011509893.2 XP_011508195.1 7 2368
          XM_011509894.2 XP_011508196.1 6 2317
          XM_011509897.1 XP_011508199.1 7 752
          XM_011509898.2 XP_011508200.1 7 683
          XM_017002081.1 XP_016857570.1 6 2312
          XM_017002082.1 XP_016857571.1 6 2313
          XM_017002083.1 XP_016857572.1 7 752
          GenBank mRNA AB208849.1 - 4 271
          AB451255.1 - 7 933
          AB451379.1 - 7 933
          AK096169.1 - 7 1137
          AK130014.1 - 6 487
          AK225332.1 - 7 1079
          AK292143.1 - 8 842
          AK296786.1 - 8 965
          AK315842.1 - 8 753
          BC014158.2 - 8 723 4541216
          BC033925.1 - 5 556 4896385
          BX647136.1 - 8 1070
          BX647149.1 - 7 1154
          U73377.1 - 7 1127
          X68148.1 - 8 684

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPK activity
          angiogenesis
          response to hypoxia
          epidermal growth factor receptor signaling pathway
          regulation of epidermal growth factor-activated receptor activity
          Ras protein signal transduction
          heart development
          aging
          positive regulation of cell proliferation
          insulin receptor signaling pathway
          response to toxic substance
          viral process
          single organismal cell-cell adhesion
          organ regeneration
          neuron projection development
          actin cytoskeleton reorganization
          response to nicotine
          IRE1-mediated unfolded protein response
          Fc-epsilon receptor signaling pathway
          ERBB2 signaling pathway
          regulation of growth
          response to hydrogen peroxide
          positive regulation of GTPase activity
          positive regulation of DNA replication
          positive regulation of vasoconstriction
          positive regulation of smooth muscle cell proliferation
          leukocyte migration
          response to glucocorticoid
          cellular response to growth factor stimulus
          phosphotyrosine binding
          transmembrane receptor protein tyrosine kinase adaptor activity
          Ras guanyl-nucleotide exchange factor activity
          epidermal growth factor receptor binding
          insulin receptor binding
          insulin-like growth factor receptor binding
          neurotrophin TRKA receptor binding
          protein binding
          phospholipid binding
          ephrin receptor binding
          protein phosphatase 2A binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0