Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281786.1 | NP_001268715.1 | 6 | 747 | ||
NM_004169.4 | NP_004160.3 | 7 | 863 | |||
NM_148918.2 | NP_683718.1 | 7 | 863 | |||
XM_005256767.3 | XP_005256824.1 | 8 | 934 | |||
XM_011523992.2 | XP_011522294.1 | 7 | 831 | |||
XM_017024957.1 | XP_016880446.1 | 7 | 688 | |||
XM_017024958.1 | XP_016880447.1 | 8 | 934 | |||
XM_017024959.1 | XP_016880448.1 | 5 | 555 | |||
XM_017024960.1 | XP_016880449.1 | 5 | 555 | |||
GenBank mRNA | AK223552.1 | - | 7 | 794 | ||
AK298415.1 | - | 6 | 679 | |||
BC007979.2 | - | 7 | 709 | 3687873 | ||
BC022874.1 | - | 7 | 713 | 4108106 | ||
BC038598.1 | - | 7 | 720 | 4523709 | ||
L11931.1 | - | 6 | 676 | |||
L23928.1 | - | 6 | 664 | |||
Y14485.1 | - | 7 | 829 | |||
Y14486.1 | - | 2 | 974 |