Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RELN SIRNA Assays ›
          Gene Symbol:
          Reln
          Gene Name:
          reelin
          Gene Aliases:
          Reelen, Rl, reeler
          Chromosome Location:
          Chr.4: 9347533 - 9774257 on Build Rnor_6.0
          UniGene ID:
          Rn.98353
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s128347

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Reln
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_080394.2 NP_536319.2 27 3916
          XM_006235873.2 XP_006235935.1 27 3990
          GenBank mRNA AB049473.1 - 27 4084
          AB062680.1 - 27 3916

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          extracellular matrix glycoprotein extracellular matrix protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis involved in differentiation
          neuron migration
          proteolysis
          cell adhesion
          axon guidance
          central nervous system development
          brain development
          midgut development
          memory
          long-term memory
          locomotory behavior
          associative learning
          response to mechanical stimulus
          response to toxic substance
          response to hormone
          glial cell differentiation
          response to lithium ion
          regulation of gene expression
          positive regulation of neuron projection development
          positive regulation of phosphatidylinositol 3-kinase signaling
          response to organic cyclic compound
          dendrite development
          cell migration
          response to insecticide
          peptidyl-tyrosine phosphorylation
          spinal cord patterning
          ventral spinal cord development
          dentate gyrus development
          hippocampus development
          orbitofrontal cortex development
          cerebral cortex tangential migration
          layer formation in cerebral cortex
          hypothalamus development
          cerebral cortex development
          forebrain development
          positive regulation of TOR signaling
          response to progesterone
          positive regulation of CREB transcription factor activity
          social behavior
          protein localization to synapse
          reelin-mediated signaling pathway
          thyroid hormone metabolic process
          response to drug
          response to amino acid
          response to ethanol
          positive regulation of protein kinase activity
          response to pain
          response to axon injury
          positive regulation of peptidyl-tyrosine phosphorylation
          regulation of behavior
          modulation of synaptic transmission
          positive regulation of small GTPase mediated signal transduction
          response to corticosterone
          response to electrical stimulus
          positive regulation of synaptic transmission, glutamatergic
          positive regulation of dendritic spine morphogenesis
          positive regulation of protein tyrosine kinase activity
          response to fatty acid
          cellular response to dexamethasone stimulus
          positive regulation of synapse maturation
          NMDA glutamate receptor clustering
          postsynaptic density protein 95 clustering
          receptor localization to synapse
          lateral motor column neuron migration
          positive regulation of long-term synaptic potentiation
          positive regulation of lateral motor column neuron migration
          regulation of N-methyl-D-aspartate selective glutamate receptor activity
          positive regulation of excitatory postsynaptic potential
          positive regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
          molecular_function
          protein serine/threonine/tyrosine kinase activity
          serine-type peptidase activity
          metal ion binding
          lipoprotein particle receptor binding
          very-low-density lipoprotein particle receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline