Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SPARC SIRNA Assays ›
Gene Symbol:
Sparc
Gene Name:
secreted protein, acidic, cysteine-rich (osteonectin)
Gene Aliases:
-
Chromosome Location:
Chr.10: 40742390 - 40764222 on Build Rnor_6.0
UniGene ID:
Rn.234316
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s128434
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
404.00
Your Price
Online offer:
391.65
391.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Sparc
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012656.1
NP_036788.1
8
576
XM_008767663.1
XP_008765885.1
7
654
GenBank mRNA
BC061777.1
-
7
591
5621666
D28875.1
-
7
593
FQ212364.1
-
7
645
FQ212499.1
-
7
610
FQ213815.1
-
7
610
FQ213945.1
-
7
610
FQ214222.1
-
7
608
FQ214492.1
-
7
610
FQ214649.1
-
7
610
FQ214673.1
-
7
610
FQ215195.1
-
7
609
FQ215581.1
-
7
611
FQ215622.1
-
7
612
FQ215818.1
-
7
621
FQ215935.1
-
7
610
FQ216823.1
-
7
612
FQ217546.1
-
7
611
FQ219866.1
-
7
592
FQ219932.1
-
8
592
FQ219983.1
-
7
610
FQ220003.1
-
7
610
FQ220043.1
-
7
608
FQ220046.1
-
7
610
FQ220186.1
-
7
608
FQ220279.1
-
5
434
FQ220287.1
-
7
612
FQ220443.1
-
7
592
FQ220507.1
-
7
618
FQ220547.1
-
7
610
FQ220572.1
-
7
610
FQ220644.1
-
7
610
FQ220827.1
-
7
610
FQ220880.1
-
7
610
FQ220901.1
-
7
610
FQ222224.1
-
7
610
FQ223025.1
-
7
610
FQ224996.1
-
6
481
FQ225033.1
-
7
612
FQ225092.1
-
7
606
FQ225200.1
-
7
610
FQ225220.1
-
7
589
FQ225609.1
-
7
610
FQ225618.1
-
7
608
FQ225644.1
-
3
277
FQ225678.1
-
7
610
FQ225688.1
-
7
610
FQ225694.1
-
7
612
FQ225722.1
-
7
609
FQ225884.1
-
7
608
FQ225914.1
-
7
612
FQ225965.1
-
7
610
FQ226299.1
-
7
592
FQ226354.1
-
7
610
FQ226360.1
-
7
610
FQ226584.1
-
7
610
FQ226599.1
-
7
611
FQ226651.1
-
7
633
FQ226673.1
-
7
591
FQ226731.1
-
7
610
FQ226746.1
-
7
608
FQ227628.1
-
7
619
FQ227644.1
-
7
610
FQ227725.1
-
7
611
FQ227727.1
-
7
590
FQ227749.1
-
7
610
FQ228032.1
-
7
590
FQ228597.1
-
7
592
FQ229172.1
-
7
610
FQ229199.1
-
7
610
FQ229230.1
-
7
610
FQ229283.1
-
7
592
FQ229299.1
-
7
593
FQ229315.1
-
7
608
FQ229316.1
-
7
610
FQ230029.1
-
7
610
FQ230086.1
-
7
608
FQ230210.1
-
7
611
FQ230381.1
-
7
610
FQ230595.1
-
7
592
FQ230619.1
-
7
610
FQ230667.1
-
7
611
FQ230705.1
-
7
608
FQ230778.1
-
7
610
FQ230864.1
-
7
610
FQ230993.1
-
7
610
FQ231067.1
-
7
610
FQ231166.1
-
7
609
FQ231444.1
-
7
609
FQ231523.1
-
7
610
FQ231543.1
-
7
610
FQ231632.1
-
7
610
FQ231753.1
-
7
607
FQ231815.1
-
7
592
FQ231918.1
-
7
592
FQ231975.1
-
7
592
FQ232042.1
-
7
609
FQ232191.1
-
7
610
FQ232201.1
-
7
610
FQ232223.1
-
7
610
FQ232271.1
-
7
607
FQ232364.1
-
7
612
FQ232445.1
-
7
608
FQ232464.1
-
7
610
FQ232710.1
-
7
608
FQ232838.1
-
7
610
FQ232847.1
-
7
610
FQ232862.1
-
7
608
FQ232869.1
-
7
610
FQ232965.1
-
7
610
FQ232977.1
-
7
610
FQ232986.1
-
7
610
FQ233178.1
-
7
610
FQ233283.1
-
7
589
FQ233669.1
-
7
609
FQ233698.1
-
7
592
FQ233777.1
-
7
608
FQ233810.1
-
7
612
FQ233845.1
-
7
610
FQ233850.1
-
7
611
FQ233926.1
-
7
610
FQ234037.1
-
7
610
FQ234069.1
-
7
610
FQ234108.1
-
7
610
FQ234199.1
-
7
610
FQ234410.1
-
7
592
FQ234418.1
-
7
610
FQ234523.1
-
7
597
FQ234589.1
-
7
608
FQ234657.1
-
7
592
FQ234685.1
-
7
610
FQ234766.1
-
7
607
FQ235008.1
-
7
612
FQ235038.1
-
7
610
FQ235049.1
-
7
592
Y13714.1
-
8
576
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009242, AK082283, CT010306, AK148670, AK146935, and others.
Panther Classification:
Molecular Function -
extracellular matrix glycoprotein
Gene Ontology Categories:
Function(s)
Process(es)
ossification
negative regulation of endothelial cell proliferation
signal transduction
heart development
response to gravity
response to lead ion
positive regulation of endothelial cell migration
negative regulation of angiogenesis
regulation of cell morphogenesis
lung development
response to lipopolysaccharide
response to L-ascorbic acid
response to cytokine
wound healing
regulation of cell proliferation
response to peptide hormone
response to ethanol
response to cadmium ion
inner ear development
response to glucocorticoid
response to cAMP
response to calcium ion
bone development
cellular response to growth factor stimulus
calcium ion binding
collagen binding
extracellular matrix binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-5mtvh:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0