Search
Search

SIX1| Gene Symbol: SIX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005982.3 | NP_005973.1 | 1 | 390 | ||
| XM_017021602.1 | XP_016877091.1 | 1 | 442 | |||
| GenBank mRNA | BC008874.2 | - | 1 | 390 | 4138234 | |
| BT007083.1 | - | 1 | 142 | |||
| X91868.1 | - | 1 | 417 | |||