Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SIX1 SIRNA Assays ›
Gene Symbol:
SIX1
Gene Name:
SIX homeobox 1
Gene Aliases:
BOS3, DFNA23, TIP39
Chromosome Location:
Chr.14: 60644698 - 60649489 on Build GRCh38
UniGene ID:
Hs.633506
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s12875
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SIX1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_005982.3
NP_005973.1
1, 2
816
GenBank mRNA
BC008874.2
-
1, 2
816
4138234
BT007083.1
-
1, 2
568
X91868.1
-
1, 2
843
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_053759, AF323957.
This siRNA also targets the following transcripts in Mouse: NM_009189, AK131925, X80339, AK144753, BC023304, and others.
Panther Classification:
Molecular Function -
homeodomain transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
ureteric bud development
branching involved in ureteric bud morphogenesis
organ induction
kidney development
outflow tract morphogenesis
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
apoptotic process
pattern specification process
skeletal muscle tissue development
sensory perception of sound
cell proliferation
regulation of synaptic growth at neuromuscular junction
regulation of skeletal muscle satellite cell proliferation
facial nerve morphogenesis
epithelial cell differentiation
thyroid gland development
olfactory placode formation
regulation of protein localization
protein localization to nucleus
aorta morphogenesis
inner ear morphogenesis
middle ear morphogenesis
negative regulation of neuron apoptotic process
regulation of neuron differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
thymus development
neuron fate specification
generation of neurons
embryonic cranial skeleton morphogenesis
embryonic skeletal system morphogenesis
inner ear development
regulation of epithelial cell proliferation
myoblast migration
pharyngeal system development
myotome development
fungiform papilla morphogenesis
trigeminal ganglion development
otic vesicle development
metanephric mesenchyme development
regulation of branch elongation involved in ureteric bud branching
positive regulation of ureteric bud formation
mesonephric tubule formation
ureter smooth muscle cell differentiation
positive regulation of secondary heart field cardioblast proliferation
cochlea morphogenesis
positive regulation of branching involved in ureteric bud morphogenesis
negative regulation of branching involved in ureteric bud morphogenesis
positive regulation of mesenchymal cell proliferation involved in ureter development
regulation of skeletal muscle cell differentiation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
sequence-specific DNA binding
transcription regulatory region DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0