Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DBI SIRNA Assays ›
          Gene Symbol:
          Dbi
          Gene Name:
          diazepam binding inhibitor (GABA receptor modulator, acyl-CoA binding protein)
          Gene Aliases:
          Acbp, Acoabp3, Ep, Odn, RNACOABP3, Ttn
          Chromosome Location:
          Chr.13: 36147667 - 36156076 on Build Rnor_6.0
          UniGene ID:
          Rn.3285
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s128869

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Dbi
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031853.4 NP_114054.1 2 229
          GenBank mRNA AY383701.1 - 409
          BC084717.1 - 1 116 7310272
          FQ209769.1 - 2 155
          FQ209835.1 - 2 177
          FQ209938.1 - 2 172
          FQ210140.1 - 2 177
          FQ210165.1 - 2 172
          FQ210336.1 - 2 174
          FQ210781.1 - 2 204
          FQ210997.1 - 2 174
          FQ211081.1 - 2 177
          FQ211136.1 - 2 177
          FQ211192.1 - 2 177
          FQ211339.1 - 2 178
          FQ211377.1 - 1 98
          FQ213527.1 - 2 178
          FQ214572.1 - 2 177
          FQ217714.1 - 2 178
          FQ218164.1 - 2 196
          FQ218239.1 - 2 177
          FQ218680.1 - 2 177
          FQ219739.1 - 2 177
          FQ221153.1 - 2 172
          FQ221871.1 - 2 178
          FQ222052.1 - 2 177
          FQ222378.1 - 2 179
          FQ222416.1 - 2 172
          FQ222549.1 - 2 164
          FQ222564.1 - 2 178
          FQ222579.1 - 2 208
          FQ223167.1 - 2 172
          FQ223413.1 - 2 177
          FQ223492.1 - 2 178
          FQ223501.1 - 2 177
          FQ223602.1 - 2 135
          FQ224361.1 - 2 177
          FQ224797.1 - 2 178
          FQ229114.1 - 2 172
          M14201.1 - 2 229
          M20268.1 - 2 118

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_007830, NM_001037999, AK145278, AK027906, AK008540, and others.

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          behavioral fear response
          hair follicle development
          acyl-CoA metabolic process
          triglyceride metabolic process
          steroid biosynthetic process
          brain development
          learning or memory
          glial cell proliferation
          protein palmitoylation
          lateral ventricle development
          pancreatic juice secretion
          cellular response to nutrient
          negative regulation of fatty acid beta-oxidation
          regulation of synaptic transmission, GABAergic
          phosphatidylcholine acyl-chain remodeling
          skin development
          positive regulation of lipid biosynthetic process
          positive regulation of release of sequestered calcium ion into cytosol
          long-term synaptic potentiation
          fatty-acyl-CoA binding
          protein binding
          lipid binding
          benzodiazepine receptor binding
          long-chain fatty acyl-CoA binding
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0