Search
Search

Hspe1| Gene Symbol: Hspe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012966.1 | NP_037098.1 | 512 | |||
| GenBank mRNA | BC058492.1 | - | 287 | 6887393 | ||
| FQ209877.1 | - | 300 | ||||
| FQ220992.1 | - | 324 | ||||
| FQ222153.1 | - | 299 | ||||
| X71429.1 | - | 258 | ||||