Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC8A3 SIRNA Assays ›
          Gene Symbol:
          SLC8A3
          Gene Name:
          solute carrier family 8 member A3
          Gene Aliases:
          NCX3
          Chromosome Location:
          Chr.14: 70044217 - 70189626 on Build GRCh38
          UniGene ID:
          Hs.337696
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s13018

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SLC8A3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130417.2 NP_001123889.1 3 453
          NM_033262.4 NP_150287.1 5 2750
          NM_058240.3 NP_489479.1 5 2747
          NM_182932.2 NP_891977.1 4 2738
          NM_182936.2 NP_891981.1 2 349
          NM_183002.2 NP_892114.1 5 2756
          NR_104122.1 3 2634
          XM_006720240.3 XP_006720303.1 4 1602
          XM_017021606.1 XP_016877095.1 5 2128
          XM_017021607.1 XP_016877096.1 5 2621
          XM_017021608.1 XP_016877097.1 4 2364
          XM_017021609.1 XP_016877098.1 4 2603
          XM_017021611.1 XP_016877100.1 3 726
          GenBank mRNA AF510501.1 - 4 2738
          AF510502.1 - 5 2756
          AF510503.1 - 3 2634
          AJ304852.1 - 4 2058
          AJ304853.1 - 4 2055
          AJ508602.1 - 3 1534
          AJ745101.1 - 2 312
          AJ745102.1 - 3 416
          AK096553.1 - 2 279
          BC036783.1 - 4 2541
          BC142969.1 - 3 442
          BC150183.1 - 3 2061
          BC150193.1 - 3 2061
          BI551720.1 - 3 452
          DA351808.1 - 3 454

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          ion transport
          mitochondrial calcium ion transport
          cellular calcium ion homeostasis
          cell communication
          learning
          memory
          regulation of skeletal muscle contraction
          telencephalon development
          sodium ion transmembrane transport
          myelination
          oligodendrocyte differentiation
          mitochondrial calcium ion homeostasis
          long-term synaptic potentiation
          calcium ion transport into cytosol
          calcium ion transmembrane transport
          cellular response to cAMP
          cellular response to hypoxia
          regulation of cardiac conduction
          calcium ion export from cell
          calcium ion import into cell
          calcium:sodium antiporter activity
          calmodulin binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0