Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KDR SIRNA Assays ›
          Gene Symbol:
          Kdr
          Gene Name:
          kinase insert domain receptor
          Gene Aliases:
          Vegfr-2
          Chromosome Location:
          Chr.14: 34727677 - 34787127 on Build Rnor_6.0
          UniGene ID:
          Rn.88869
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s130201

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Kdr
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_013062.1 NP_037194.1 5 875
          GenBank mRNA BC087029.1 - 5 851 7124961
          U93306.1 - 5 875
          U93307.1 - 5 875

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          ovarian follicle development
          vasculogenesis
          response to hypoxia
          positive regulation of protein phosphorylation
          positive regulation of endothelial cell proliferation
          lymph vessel development
          cell migration involved in sprouting angiogenesis
          positive regulation of mesenchymal cell proliferation
          endocardium development
          endothelium development
          endochondral bone growth
          positive regulation of cytosolic calcium ion concentration
          aging
          positive regulation of cell proliferation
          regulation of cell shape
          male gonad development
          positive regulation of endothelial cell migration
          positive regulation of phosphatidylinositol 3-kinase signaling
          cell migration
          peptidyl-tyrosine phosphorylation
          hemopoiesis
          hyaluronan metabolic process
          lung development
          positive regulation of cell migration
          positive regulation of BMP signaling pathway
          positive regulation of TOR signaling
          embryonic hemopoiesis
          calcium-mediated signaling using intracellular calcium source
          cellular response to vascular endothelial growth factor stimulus
          peptidyl-tyrosine autophosphorylation
          vascular endothelial growth factor signaling pathway
          response to drug
          negative regulation of apoptotic process
          surfactant homeostasis
          positive regulation of MAPK cascade
          negative regulation of neuron apoptotic process
          cell fate commitment
          endothelial cell differentiation
          regulation of endothelial cell differentiation
          positive regulation of angiogenesis
          positive regulation of vasodilation
          protein autophosphorylation
          vascular endothelial growth factor receptor signaling pathway
          positive regulation of long-term neuronal synaptic plasticity
          lung alveolus development
          cell maturation
          branching morphogenesis of an epithelial tube
          neuron projection morphogenesis
          positive regulation of epithelial cell proliferation
          positive regulation of neurogenesis
          positive regulation of calcium-mediated signaling
          positive regulation of positive chemotaxis
          positive regulation of nitric-oxide synthase biosynthetic process
          positive regulation of focal adhesion assembly
          calcium ion homeostasis
          positive regulation of ERK1 and ERK2 cascade
          response to oxygen levels
          regulation of hematopoietic progenitor cell differentiation
          negative regulation of endothelial cell apoptotic process
          positive regulation of vasculogenesis
          protein tyrosine kinase activity
          vascular endothelial growth factor-activated receptor activity
          integrin binding
          protein binding
          ATP binding
          growth factor binding
          vascular endothelial growth factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline