Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KDR SIRNA Assays ›
Gene Symbol:
Kdr
Gene Name:
kinase insert domain receptor
Gene Aliases:
Vegfr-2
Chromosome Location:
Chr.14: 34727677 - 34787127 on Build Rnor_6.0
UniGene ID:
Rn.88869
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s130202
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Kdr
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_013062.1
NP_037194.1
30
4166
GenBank mRNA
BC087029.1
-
30
4142
7124961
U93306.1
-
30
4166
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
ovarian follicle development
vasculogenesis
response to hypoxia
positive regulation of protein phosphorylation
positive regulation of endothelial cell proliferation
lymph vessel development
cell migration involved in sprouting angiogenesis
positive regulation of mesenchymal cell proliferation
endocardium development
endothelium development
endochondral bone growth
positive regulation of cytosolic calcium ion concentration
aging
positive regulation of cell proliferation
regulation of cell shape
male gonad development
positive regulation of endothelial cell migration
positive regulation of phosphatidylinositol 3-kinase signaling
cell migration
peptidyl-tyrosine phosphorylation
hemopoiesis
hyaluronan metabolic process
lung development
positive regulation of cell migration
positive regulation of BMP signaling pathway
positive regulation of TOR signaling
embryonic hemopoiesis
calcium-mediated signaling using intracellular calcium source
cellular response to vascular endothelial growth factor stimulus
peptidyl-tyrosine autophosphorylation
vascular endothelial growth factor signaling pathway
response to drug
negative regulation of apoptotic process
surfactant homeostasis
positive regulation of MAPK cascade
negative regulation of neuron apoptotic process
cell fate commitment
endothelial cell differentiation
regulation of endothelial cell differentiation
positive regulation of angiogenesis
positive regulation of vasodilation
protein autophosphorylation
vascular endothelial growth factor receptor signaling pathway
positive regulation of long-term neuronal synaptic plasticity
lung alveolus development
cell maturation
branching morphogenesis of an epithelial tube
neuron projection morphogenesis
positive regulation of epithelial cell proliferation
positive regulation of neurogenesis
positive regulation of calcium-mediated signaling
positive regulation of positive chemotaxis
positive regulation of nitric-oxide synthase biosynthetic process
positive regulation of focal adhesion assembly
calcium ion homeostasis
positive regulation of ERK1 and ERK2 cascade
response to oxygen levels
regulation of hematopoietic progenitor cell differentiation
negative regulation of endothelial cell apoptotic process
positive regulation of vasculogenesis
protein tyrosine kinase activity
vascular endothelial growth factor-activated receptor activity
integrin binding
protein binding
ATP binding
growth factor binding
vascular endothelial growth factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-wq6vw:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline