Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SMARCA4 SIRNA Assays ›
          Gene Symbol:
          SMARCA4
          Gene Name:
          SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4
          Gene Aliases:
          BAF190, BAF190A, BRG1, CSS4, MRD16, RTPS2, SNF2, SNF2L4, SNF2LB, SWI2, hSNF2b
          Chromosome Location:
          Chr.19: 10960922 - 11062282 on Build GRCh38
          UniGene ID:
          Hs.327527
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s13140

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SMARCA4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001128844.1 NP_001122316.1 25, 26 3742
          NM_001128845.1 NP_001122317.1 23, 24 3382
          NM_001128846.1 NP_001122318.1 23, 24 3382
          NM_001128847.1 NP_001122319.1 23, 24 3382
          NM_001128848.1 NP_001122320.1 23, 24 3382
          NM_001128849.1 NP_001122321.1 24, 25 3456
          NM_003072.3 NP_003063.2 24, 25 3666
          XM_006722845.2 XP_006722908.1 24, 25 3588
          XM_006722846.2 XP_006722909.1 24, 25 3682
          XM_011528198.1 XP_011526500.1 25, 26 3663
          XM_017027160.1 XP_016882649.1 25, 26 3663
          XM_017027161.1 XP_016882650.1 25, 26 3663
          XM_017027162.1 XP_016882651.1 25, 26 3663
          XM_017027163.1 XP_016882652.1 25, 26 3663
          XM_017027164.1 XP_016882653.1 25, 26 3663
          XM_017027165.1 XP_016882654.1 25, 26 3663
          XM_017027166.1 XP_016882655.1 25, 26 3663
          XM_017027167.1 XP_016882656.1 25, 26 3663
          XM_017027168.1 XP_016882657.1 25, 26 3663
          GenBank mRNA AB209313.2 - 23, 24 3256
          AK026573.1 - 6, 7 761
          AK055168.1 - 7, 8 1454
          AK097105.1 - 6, 7 1911
          AK290033.1 - 25, 26 3663
          AK299761.1 - 10, 11 1124
          AK303352.1 - 9, 10 1066
          AK308763.1 - 9, 10 1295
          BC136644.1 - 22, 23 3602 9020634
          BC150298.1 - 24, 25 3587 8860365
          D26156.1 - 24, 25 3439
          EU430756.1 - 23, 24 3382
          EU430757.1 - 23, 24 3382
          EU430758.1 - 23, 24 3382
          EU430759.1 - 23, 24 3382
          U29175.1 - 24, 25 3460

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA helicase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          neural retina development
          nucleosome disassembly
          chromatin remodeling
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          negative regulation of transcription from RNA polymerase II promoter during mitosis
          spermatid development
          nervous system development
          covalent chromatin modification
          positive regulation of Wnt signaling pathway
          negative regulation of cell growth
          ATP-dependent chromatin remodeling
          positive regulation by host of viral transcription
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of androgen receptor signaling pathway
          positive regulation of transcription of nuclear large rRNA transcript from RNA polymerase I promoter
          positive regulation of glucose mediated signaling pathway
          positive regulation of pri-miRNA transcription from RNA polymerase II promoter
          beta-catenin-TCF complex assembly
          negative regulation of G1/S transition of mitotic cell cycle
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          RNA polymerase II transcription coactivator activity
          RNA polymerase I CORE element sequence-specific DNA binding
          p53 binding
          transcription coactivator activity
          transcription corepressor activity
          helicase activity
          protein binding
          ATP binding
          DNA-dependent ATPase activity
          transcription factor binding
          Tat protein binding
          nucleosomal DNA binding
          protein N-terminus binding
          androgen receptor binding
          DNA polymerase binding
          lysine-acetylated histone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0