Search
Search

Grk6| Gene Symbol: Grk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112712.1 | NP_001106183.1 | 14 | 1499 | ||
| NM_001112713.1 | NP_001106184.1 | 14 | 1499 | |||
| NM_031657.3 | NP_113845.2 | 14 | 1499 | |||
| XM_006253653.2 | XP_006253715.1 | 14 | 1608 | |||
| XM_008771504.1 | XP_008769726.1 | 13 | 1766 | |||
| GenBank mRNA | AF040750.1 | - | 14 | 1499 | ||
| X97439.1 | - | 14 | 1499 | |||
| X97440.1 | - | 14 | 1499 | |||
| Y09365.1 | - | 14 | 1527 | |||