Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GHRL SIRNA Assays ›
Gene Symbol:
Ghrl
Gene Name:
ghrelin/obestatin prepropeptide
Gene Aliases:
-
Chromosome Location:
Chr.4: 145674157 - 145678066 on Build Rnor_6.0
UniGene ID:
Rn.42103
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s133177
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ghrl
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_021669.2
NP_067701.1
3
332
GenBank mRNA
AB029433.1
-
3
333
AB035699.1
-
3
326
Back To Top
More Information
Panther Classification:
Molecular Function -
peptide hormone
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
gastric acid secretion
negative regulation of endothelial cell proliferation
G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
actin polymerization or depolymerization
adult feeding behavior
response to hormone
hormone-mediated signaling pathway
dendrite development
positive regulation of insulin secretion
regulation of response to food
positive regulation of response to food
positive regulation of appetite
negative regulation of interleukin-1 beta production
negative regulation of locomotion
regulation of cell proliferation
negative regulation of circadian sleep/wake cycle, REM sleep
negative regulation of tumor necrosis factor biosynthetic process
negative regulation of apoptotic process
response to estrogen
negative regulation of interleukin-6 biosynthetic process
positive regulation of circadian sleep/wake cycle, non-REM sleep
negative regulation of insulin secretion
decidualization
negative regulation of inflammatory response
positive regulation of corticotropin secretion
positive regulation of cortisol secretion
positive regulation of synapse assembly
excitatory postsynaptic potential
positive regulation of growth hormone secretion
positive regulation of protein tyrosine kinase activity
negative regulation of energy homeostasis
positive regulation of energy homeostasis
G-protein coupled receptor binding
hormone activity
growth hormone-releasing hormone activity
protein tyrosine kinase activator activity
ghrelin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0